Informational:An new endotoxin detection kit is now available—Pierce Chromogenic Endotoxin Quant Kit, with higher sensitivity and no interference from β-glucans. To learn more about these new kits, please visit thermofisher.com/endotoxin.
Thermo Scientific™

Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL

Catalog number: 88273
Have Questions?
Thermo Scientific™

Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.25 mL

Catalog number: 88273
Have Questions?
Catalog Number
88273
Unit Size
5 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
882735 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce High Capacity Endotoxin Removal Resin binds to reduce endotoxin levels in protein samples by ≥99% in 1 hour; with this spin cup format, protein recovery is ≥85%.

Features of High Capacity Endotoxin Removal Resin Spin Columns:

High capacity—bind up to 2,000,000 EU/mL to eliminate >99% of endotoxins
Selective—recover ≥85% of your protein sample
High performance—meets FDA guidelines by reducing final EU concentration to <5 EU/mL
Fast—our new spin cup format enables endotoxin depletion within 1 hour
Clean—single-use spin columns avoid cross contamination of samples
Optimized—spin cups are optimized for different sample volumes

The Pierce High Capacity Endotoxin Removal Resin combines porous cellulose beads and an FDA-approved food preservative, poly(ε-lysine), as an affinity ligand to selectively bind endotoxins. The modified polylysine affinity ligand eliminates the toxicity concerns associated with alternative technologies using polymixin B ligands and sodium deoxycholate buffers.

Requires:
Endotoxin-free water
Applications:

Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection

The presence of endotoxins in biologically derived products prepared for research, vaccination, or therapeutic use is a major concern due to the diverse and potentially harmful biological response to these molecules. Endotoxin contamination in proteins purified from bacterial cultures is due to residual lipopolysaccharides (LPS) derived from the outer cell membrane of gram-negative bacteria such as E. coli. Endotoxin-contaminated protein samples transfected into cells or injected into an animal host can initiate a strong immune response resulting in a Systemic Inflammatory Response Syndrome (SIRS) and/or sepsis. Traditional endotoxin removal resins that use polymixin B and sodium deoxycholate to clear endotoxins from a sample also have the negative side effects of neurotoxicity and acute renal tubular necrosis.

Pierce High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000-EU/mL binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/mL. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/mL. This spin cup format is a fast, single-use method to remove endotoxins from protein samples in as little as 1 hour. The spin cups use a batch format to bind and remove endotoxins while allowing for >85% protein recovery.

One endotoxin unit/mL (EU/mL) equals approximately 0.1ng endotoxin/mL of solution.

More Product Data
Removing endotoxins using a spin-column format
Eliminate endotoxins from protein and antibody samples

Related Products
Pierce™ High Capacity Endotoxin Removal Resin
Pierce™ High Capacity Endotoxin Removal Spin Columns, 0.5 mL
Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
Up to 2,000,000 EU/mL
Formulation
5mL-capacity centrifuge columns containing 0.25mL resin
Form
Liquid
Product Line
Pierce™
Quantity
5 Columns
Target
Endotoxin Removal
Type
Spin Column
Stationary Phase
Proprietary Ligand
Format
Spin Column
Column Type
Affinity, Cellulose Resin

Contents & Storage

Upon receipt store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text