Thermo Scientific™

Pierce™ Primary Cardiomyocyte Isolation Kit

Catalog number: 88281
Have Questions?
Thermo Scientific™

Pierce™ Primary Cardiomyocyte Isolation Kit

Catalog number: 88281
Have Questions?
Catalog Number
88281
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
882811 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Primary Cardiomyocyte Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary cardiomyocytes from neonatal heart from mouse and rat.

Features of the Primary Cardiomyocyte Isolation Kit:

Optimized – procedure and reagents optimized for viability, yield, purity, and ease-of-use
Time-saving – isolation protocol requires less than 2 hours compared to up to 21 hours by other methods
Yield – provides a 1.5- to 4-fold increase in yield compared to do-it-yourself methods and other commercial kits
Viability – provides higher viability than do-it-yourself methods and other commercial kits
Functional – cultured cardiomyocytes express the appropriate biochemical markers with validated beating activity

The Primary Cardiomyocyte Isolation Kit has been designed to obtain high viability and purity of primary cardiomyocytes using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains two tissue-specific dissociation enzymes, media formulated for primary cell culture, and a cardiomyocyte cell culture growth supplement that promotes the isolation, enrichment and growth of functional cardiomyocytes. The validated protocol ensures optimal isolation and significantly reduces the processing time of isolation when compared to other methods. Addition of the cardiomyocyte growth supplement to culture cells further enriches for cardiomycytes, and cultures have been maintained for up to 7 days using the kit components.

Includes:
• Complete kit includes Cardiomyocyte Isolation Enzyme 1, Cardiomyocyte Isolation Enzyme 2, Hanks' Balanced Salt Solution (HBSS), DMEM for Primary Cell Isolation, and Cardiomyocyte Growth Supplement

Applications:
• Cardiomyocyte cell differentiation
• Immunohistochemistry (IHC)
• Functional and biochemical assays
• Biologically relevant system for preclinical drug discovery and predictive disease modeling

Current methods for dissociation of cardiac cells from heart tissue require repeated (5 to 8 incubations) or lengthy (>16 hours) enzyme digestion, resulting in reduced yield and viability of isolated cells. The Pierce Primary Cardiomyocyte Isolation Kit provides fully optimized reagents and a protocol that prevents overdigestion for the isolation and culture of cardiomyocytes. The complete process from handling primary tissues to seeding cells in culture vessels can be completed within 2 hours.

Because it uses optimized tissue-specific dissociation enzymes, this kit isolates cardiomyocytes with greater yield and cell viability than cells isolated by existing methods. The isolated primary cardiomyocytes express cardiomyocyte protein markers and exhibit contractile function (beating activity, see video), thereby providing a model system for studies of contraction, ischaemia, hypoxia and the toxicity of various compounds.

   

More Product Data
One-hour procedure to isolate primary cardiomyocytes from neonatal mouse and rat hearts

Related Products
Cardiomyocyte Isolation Enzyme 1 (with papain) for Pierce™ Primary Cell Isolation Kits
Cardiomyocyte Isolation Enzyme 2 (with thermolysin) (20X) for Pierce™ Primary Cell Isolation Kits
DMEM for Pierce™ Primary Cell Isolation Kits
Hanks' Balanced Salt Solution (HBSS) without Ca2+, Mg2+ for Pierce™ Primary Cell Isolation Kits
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Primary Cell Isolation Kit
Cell Type
Cardiomyocytes, Primary Cells
Target Species
Rat, Mouse
Product Line
Pierce™
For Use With (Application)
Immunohistochemistry
Quantity
1 Kit

Contents & Storage

Upon receipt store Product 88281X at 4°C and Product 88281Y at -20°C. Product 88281X is shipped on ice packs and Product 88281Y is shipped with dry ice.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text