Cardiomyocyte Isolation Enzyme 2 (with thermolysin) is a tissue-specific dissociation reagent for the isolation of primary cardiomyocytes from neonatal heart tissue from mouse/rat and is a component of the Thermo Scientific™ Pierce™ Primary Cardiomyocyte Isolation Kit (Product No. 88281).
Thermolysin is a thermostable extracellular metalloendopeptidase. When used with Cardiomyocyte Isolation Enzyme 1 (with papain), it improves digestion efficiency and cell yield of primary cardiomyocytes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
Cell Isolation Kit
Cell Type
Cardiomyocytes, Primary Cells
Target Species
Rat, Mouse
Form
Solid
Product Line
Pierce™
Quantity
1 Vial
Contents & Storage
Stability: 1 year at -20°C Enzyme performance is not affected for up to two freeze-thaw cycles. Diluted enzyme is not stable and not recommended for long-term storage.