Pierce™ Triple Quadrupole Calibration Solution
Pierce™ Triple Quadrupole Calibration Solution
Thermo Scientific™

Pierce™ Triple Quadrupole Calibration Solution

Thermo Scientific Pierce Triple Quadrupole Calibration Solution is a mixture of high purity, ionizable components specifically designed for positive modeRead more
Have Questions?
Catalog number 88325
Price (USD)/ 10 mL
264.65
Online exclusive
289.00 
Save 24.35 (8%)
-
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (USD)/ 10 mL
264.65
Online exclusive
289.00 
Save 24.35 (8%)
Add to cart
Pierce™ Triple Quadrupole Calibration Solution
Catalog number88325
Price (USD)/ 10 mL
264.65
Online exclusive
289.00 
Save 24.35 (8%)
-
Add to cart
Thermo Scientific Pierce Triple Quadrupole Calibration Solution is a mixture of high purity, ionizable components specifically designed for positive mode calibration of Thermo Scientific Triple Stage Quadrupole instruments.

Features of Triple Quadrupole Calibration Solution:

Strong peaks—mixture of three tyrosine polymers in methanol/formic acid solution
Ready-to-use—load the reference standard into a syringe and inject into the instrument
High-purity—mass spectrometry grade reagents in a non-leachable container
Stable—store at 2 to 8°C for more than one year (see CofA for specific expiration date of each lot)

Pierce Triple Quadrupole Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the required routine calibration to maintain the robust performance of Thermo Scientific Mass Spectrometers. The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

Pierce Triple Quadrupole Calibration Solution can be used to calibrate the TSQ Quantum series, TSQ Discovery series, TSQ Quantum Access Series and TSQ Vantage Series Mass Spectrometer instruments.

More Product Data
Calibration solutions for Thermo Scientific Mass Spectrometers
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Mass Spectrometer
Product TypeCalibration Solution
Product LinePierce™
Starting MaterialSmall Molecules
Final Product TypeSmall Molecules
Detection MethodMass Spectrometry
Workflow StepCalibration
Quantity10 mL
FormLiquid
Unit Size10 mL
Contents & Storage
Store between 2°C and 8°C.
1x1 image pixel for data collection