Thermo Scientific™

Pierce™ 6 Protein Digest, equimolar, LC-MS grade

Catalog number: 88342
Have Questions?
Thermo Scientific™

Pierce™ 6 Protein Digest, equimolar, LC-MS grade

Catalog number: 88342
Have Questions?
Catalog Number
88342
Unit Size
100 pmol
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
88342100 pmol
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ Pierce™ 6 Protein Digest Standard, Equimolar, LC-MS Grade, is a highly validated equimolar protein digest mixture for use as a quality control sample for mass spectrometry (MS) analysis of proteomic samples.

Positive control sample—optimized and validated as a quality control standard for mass spectrometry applications
Excellent sequence coverage—on average, greater than 70% sequence coverage of the 6-protein mixture
Validated peptide quality—digestion procedure optimized for minimal missed cleavages and over-alkylation
Rigorously tested—high-quality, consistent, 6-protein digest guaranteed and documented via lot-specific certificates of analysis
Stable—provided in a stable lyophilized format

The Pierce 6 Protein Digest Standard contains an equimolar mixture of highly pure bovine cytochrome c, lysozyme, alcohol dehydrogenase, bovine serum albumin, apo-transferrin, and beta-galactosidase. The proteins are reduced, alkylated (iodoacetic acid), digested (MS-grade trypsin), and desalted (C18) after trypsin digestion to provide a quality LC/MS-grade standard free of intact protein. The peptide digest provides excellent sequence coverage with minimal over-alkylation and missed cleavages. This digest has been robustly tested for peptide quality, digestion efficiency, and lot-to-lot uniformity and is suitable for LC and LC-MS applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Mass Spectrometer
Product Type
Protein Digest
Product Line
Pierce™
Starting Material
Peptides
Final Product Type
Peptides
Detection Method
Mass Spectrometry
Workflow Step
Calibration
Form
Solid

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text