Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product Type | Cup, Conical |
Product Line | Slide-A-Lyzer™ |
MWCO | 3.5 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.5 mL |
Sufficient For | 25 Samples, 50–500 μL each |
Quantity | 25 Devices |
Unit Size | 25 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 3.5 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Sufficient For | 50 Samples, 10–100 μL each |
Quantity | 50 Devices |
Unit Size | 50 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 3.5 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Sufficient For | 250 Samples, 10–100 μL each |
Quantity | 250 Devices |
Unit Size | 250 devices |
Product Type | Cup, Microcentrifuge Kit |
Product Line | Slide-A-Lyzer™ |
MWCO | 3.5 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device Kit |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Sufficient For | 10 Samples, 10–100 μL each |
Quantity | 10 Devices |
Unit Size | 10 devices |
Product Type | Cup, Conical |
Product Line | Slide-A-Lyzer™ |
MWCO | 3.5 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Description | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 2 mL |
Sufficient For | 25 Samples, 200–2000 μL each |
Quantity | 25 Devices |
Unit Size | 25 devices |
Catalog Number | Specifications | Unit Size | Product Type | Sample Volume (Metric) | Sufficient For | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
88400 | 25 devices | Cup, Conical | 0.5 mL | 25 Samples, 50–500 μL each | 269.65 Online exclusive 289.00 Save 19.35 (7%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69550 | 50 devices | Cup, Microcentrifuge | 0.1 mL | 50 Samples, 10–100 μL each | 229.65 Online exclusive 246.00 Save 16.35 (7%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69552 | 250 devices | Cup, Microcentrifuge | 0.1 mL | 250 Samples, 10–100 μL each | 706.65 Online exclusive 760.00 Save 53.35 (7%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69558 | 10 devices | Cup, Microcentrifuge Kit | 0.1 mL | 10 Samples, 10–100 μL each | 105.65 Online exclusive 113.00 Save 7.35 (7%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88403 | 25 devices | Cup, Conical | 2 mL | 25 Samples, 200–2000 μL each | 293.65 Online exclusive 315.00 Save 21.35 (7%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The 0.5mL and 2mL MINI Dialysis Devices fit in and are capped by inserting them into the included 15mL and 50mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI Dialysis Devices, low molecular weight contaminant removal, buffer exchange and desalting, can be accomplished within 4 to 8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.