Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product Type | Cup, 15 mL Conical |
Product Line | Slide-A-Lyzer™ |
MWCO | 20 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.5 mL |
Sufficient For | 25 Samples, 50–500 μL each |
Quantity | 25 Devices |
Unit Size | 25 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 20 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Sufficient For | 50 Samples, 10–100 μL each |
Quantity | 50 Devices |
Unit Size | 50 devices |
Product Type | Cup, Microcentrifuge |
Product Line | Slide-A-Lyzer™ |
MWCO | 20 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 0.1 mL |
Sufficient For | 250 Samples, 10–100 μL each |
Quantity | 250 Devices |
Unit Size | 250 devices |
Product Type | Cup, 50 mL Conical |
Product Line | Slide-A-Lyzer™ |
MWCO | 20 kDa |
Membrane | Cellulose Membrane, Dialysis |
Format | MINI Dialysis Device |
Purification Target | Buffer Exchange, Protein |
Sample Volume (Metric) | 2 mL |
Sufficient For | 25 Samples, 200–2000 μL each |
Quantity | 25 Devices |
Unit Size | 25 devices |
Catalog Number | Specifications | Unit Size | Product Type | Sample Volume (Metric) | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
88402 | 25 devices | Cup, 15 mL Conical | 0.5 mL | 25 Devices | 266.65 Online exclusive 289.00 Save 22.35 (8%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69590 | 50 devices | Cup, Microcentrifuge | 0.1 mL | 50 Devices | 225.65 Online exclusive 244.00 Save 18.35 (8%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
69555 | 250 devices | Cup, Microcentrifuge | 0.1 mL | 250 Devices | 699.65 Online exclusive 760.00 Save 60.35 (8%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88405 | 25 devices | Cup, 50 mL Conical | 2 mL | 25 Devices | 289.65 Online exclusive 313.00 Save 23.35 (7%) | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The 0.5mL and 2mL MINI Dialysis Devices fit in and are capped by inserting them into the included 15mL and 50mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI Dialysis Devices, low molecular weight contaminant removal, buffer exchange and desalting, can be accomplished within 4 to 8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.
The 0.1mL MINI Dialysis Devices are designed to be capped (caps included) and inserted into standard 1.5mL microcentrifuge tubes. Alternatively, as many as 25 MINI Devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple inside a large beaker. Using the 0.1mL MINI Device, 100μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes.