PVDF Transfer Membranes, 0.2 μm, 1 Roll
PVDF Transfer Membranes, 0.2 μm, 1 Roll
Thermo Scientific™

PVDF Transfer Membranes, 0.2 μm, 1 Roll

Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods.
Have Questions?
Catalog NumberQuantity
885201 Roll
Catalog number 88520
Price (USD)
490.65
Online Exclusive
556.00
Save 65.35 (12%)
Each
-
Add to cart
Quantity:
1 Roll
Request bulk or custom format
Price (USD)
490.65
Online Exclusive
556.00
Save 65.35 (12%)
Each
Add to cart
Ask our AI about this Product
Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods. The 0.2-μm pore size is ideal for protein analysis of small amounts of proteins (as little as 10 pmoles), amino acid analysis, and transfer of small molecular weight proteins and peptides. For best results, preactivate the membrane with 100% alcohol.

Features include:
• Re-probe characteristics—high mechanical strength makes PVDF an excellent membrane for stripping and re-probing
• Durability—compatible with most organic solvents, acids, and mild bases
• Compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling. Choose Low-Fluorescence PVDF Transfer Membrane for fluorescent western blotting
• High sensitivity—exceptional binding capacity makes PVDF the ultimate choice for low abundance proteins

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity

Goat IgG: 448 μg/cm2
BSA: 340 μg/cm2
Insulin: 262 μg/cm2

Product TypePVDF Transfer Membrane
Quantity1 Roll
Shipping ConditionRoom Temperature
Dimensions (LxW)3.75 m x 26.5 cm
FormatRoll
Length (Metric)3.75 m
MaterialPVDF
Pore Size0.2 μm
Sufficient For≥100 Mini-gel Blots (When cut to 8.8 x 10 cm); ≥80 Midi-gel Blots (When cut to 8.5 x 13.25 cm)
Width (Metric)26.5 cm
Unit SizeEach
Contents & Storage
Store membranes flat at ambient temperature and away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I performed a western transfer onto a PVDF membrane and the transfer efficiency was very poor. Can you offer some tips?

Here are possible causes and solutions:

- The membrane may not be properly treated prior to transfer: Make sure that the membrane is pre-wetted with a polar organic solvent such as methanol or ethanol.
- There may be poor gel to membrane contact: Ensure that the filter paper and blotting pads are well saturated with transfer buffer, taking care to remove any bubbles during the assembly of the membrane sandwich. The gel/membrane sandwich must fit securely in the two halves of the blot module. Try adding another pad or replace any pads that have lost their resiliency with fresh ones.
- Over-compression of the gel: A good indication of over-compression is if the gel has been excessively flattened. In the event that the sandwich is over-compressed, remove enough pads so that the blotter can be closed without exerting excess pressure on the gel and the membrane.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Are your PVDF and nitrocellulose membranes compatible with the Li-COR instrument?

Yes, both our PVDF and nitrocellulose membranes are compatible with the Li-COR instrument.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am planning on doing a dot blot and my sample contains acetonitrile. Can your PVDF or nitrocellulose membrane withstand acetonitrile?

Our PVDF can tolerate acetonitrile but our nitrocellulose cannot.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How can I reduce background on my PVDF western blots?

PVDF membranes require more stringent blocking steps. This can be achieved by increasing the concentration of the blocking agent 2-5 fold, increasing the blocking time, and performing the procedure at 37 degrees C. Blocking agents bind to unoccupied sites to prevent background staining and also to membrane-bound proteins, reducing non-specific interactions with the primary antibody. Examples of blocking agents are nonfat dry milk, BSA, and Casein.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link