Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Pierce™ Protein Concentrators PES, 50K MWCO, 2-6 mL |
Filter Type | Membrane Filter |
Format | 15 mL Conical, Spin Column |
Material (Membrane) | PES |
MWCO | 50 kDa |
Sample Volume | 2 to 6 mL |
Max. RCF | Swinging-bucket rotor: 4,000 x g, Fixed-angle rotor: 10,000 x g |
Product Line | Pierce™ |
Quantity | 24 Concentrators |
Shipping Condition | Room Temperature |
Type | Protein Concentrator |
Unit Size | 24 concentrators |
Description | Pierce™ Protein Concentrators PES, 50K MWCO, 0.5 mL |
Filter Type | Membrane Filter |
Format | Microcentrifuge, Spin Column |
Material (Membrane) | PES |
MWCO | 50 kDa |
Sample Volume | ≤0.5 mL |
Max. RCF | 15,000 x g |
Product Line | Pierce™ |
Quantity | 25 Concentrators |
Shipping Condition | Room Temperature |
Type | Protein Concentrator |
Unit Size | 25 concentrators |
Description | Pierce™ Protein Concentrators PES, 50K MWCO, 2-6 mL |
Filter Type | Membrane Filter |
Format | 15 mL Conical, Spin Column |
Material (Membrane) | PES |
MWCO | 50 kDa |
Sample Volume | 2 to 6 mL |
Max. RCF | Swinging-bucket rotor: 4,000 x g, Fixed-angle rotor: 10,000 x g |
Product Line | Pierce™ |
Quantity | 10 Concentrators |
Shipping Condition | Room Temperature |
Type | Protein Concentrator |
Unit Size | 10 concentrators |
Description | Pierce™ Protein Concentrators PES, 50K MWCO, 5-20 mL |
Filter Type | Membrane Filter |
Format | 50 mL Conical, Spin Column |
Material (Membrane) | PES |
MWCO | 50 kDa |
Sample Volume | 5 to 20 mL |
Max. RCF | Swinging-bucket rotor: 5,000 x g, Fixed-angle rotor: 8,000 x g |
Product Line | Pierce™ |
Quantity | 10 Concentrators |
Shipping Condition | Room Temperature |
Type | Protein Concentrator |
Unit Size | 10 concentrators |
Description | Pierce™ Protein Concentrators PES, 50K MWCO, 5-20 mL |
Filter Type | Membrane Filter |
Format | 50 mL Conical, Spin Column |
Material (Membrane) | PES |
MWCO | 50 kDa |
Sample Volume | 5 to 20 mL |
Max. RCF | Swinging-bucket rotor: 5,000 x g, Fixed-angle rotor: 8,000 x g |
Product Line | Pierce™ |
Quantity | 24 Concentrators |
Shipping Condition | Room Temperature |
Type | Protein Concentrator |
Unit Size | 24 concentrators |
Description | Pierce™ Protein Concentrators PES, 50K MWCO, 20-100 mL |
Filter Type | Membrane Filter |
Format | Spin Column |
Material (Membrane) | PES |
MWCO | 50 kDa |
Sample Volume | 20 to 100 mL |
Max. RCF | Swinging-bucket rotor: 2,000 x g, Fixed-angle rotor: not recommended |
Product Line | Pierce™ |
Quantity | 4 Concentrators |
Shipping Condition | Room Temperature |
Type | Protein Concentrator |
Unit Size | 4 concentrators |
Catalog Number | Specifications | Unit Size | Sample Volume | Quantity | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
88539 | 24 concentrators | 2 to 6 mL | 24 Concentrators | 167.65 Online price 172.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88504 | 25 concentrators | ≤0.5 mL | 25 Concentrators | 130.65 Online price 136.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88538 | 10 concentrators | 2 to 6 mL | 10 Concentrators | 79.65 Online price 86.75 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88540 | 10 concentrators | 5 to 20 mL | 10 Concentrators | 157.65 Online price 171.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88541 | 24 concentrators | 5 to 20 mL | 24 Concentrators | 318.65 Online price 344.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88542 | 4 concentrators | 20 to 100 mL | 4 Concentrators | 186.65 Online price 200.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|