1x1 image pixel for data collection
Pierce™ Protein A/G Magnetic Beads
Pierce™ Protein A/G Magnetic Beads
Thermo Scientific™

Pierce™ Protein A/G Magnetic Beads

Immunoprecipitate or affinity purify antibodies (IgG) using Protein A/G coated magnetic particles for manual or robotic magnetic separation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
888021 mL
888035 mL
Catalog number 88802
Price (USD)
239.65
Online Exclusive
272.00
Save 32.35 (12%)
Each
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (USD)
239.65
Online Exclusive
272.00
Save 32.35 (12%)
Each
Add to cart

Thermo Scientific Pierce Protein A/G Magnetic Beads are high-performance affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.

Features of Protein A/G Magnetic Beads:

  • High capacity—nearly four times higher binding capacity than typical magnetic beads from other suppliers, allowing the use of smaller amounts per experiment
  • Low non-specific binding—stable, pre-blocked beads provide clean purification products (e.g., antigen eluted in IP with antibody is devoid of contaminating proteins from complex IP matrix)
  • Flexibility—convenience of IgG binding domains of both Protein A and Protein G on one bead
  • Compatibility—beads are compatible with manual and automated applications (e.g., Thermo Scientific KingFisher Instruments)
  • Assay consistency—magnetic beads eliminate resin loss and provide for more efficient separation of solutions than traditional IP methods that use only microcentrifuge tubes

These magnetic beads are coated with genetically engineered Pierce Protein A/G, a recombinant fusion protein which combines the IgG binding domains of both Protein A and Protein G. This enables capture of antibodies from a wider range of species and isotypes than either protein alone. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in your immunoprecipitated sample. These beads can be used both manually with a magnetic stand as well as with automated platforms such as the Thermo Scientific KingFisher Instruments.

Applications:

  • IP and Co-IP experiments (see complete kit)
  • Immunoprecipitation for analysis in non-reducing conditions
  • Antibody purification

The recombinant Protein A/G that is immobilized onto the Pierce Magnetic Beads is a fusion of the IgG binding domains of both Protein A and Protein G. Protein A/G contains four Fc-binding domains from Protein A and two from Protein G, making it a convenient tool for investigating and purifying immunoglobulins. Thus, Pierce Magnetic Particles are not simply a mixed immobilization of separate Protein A and Protein G polypeptides, nor are they a mixture of Protein A magnetic beads and Protein G magnetic beads.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConcentrationSlurry: 10mg/mL, 1% solids
Ligand TypeProtein A/G
Protein FormRecombinant
Quantity1 mL
TargetAntibody
Capacity (Metric)1 mL
FormLiquid
Particle Size1 μm
Product LinePierce™
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. Product shipped with an ice pack.

Frequently asked questions (FAQs)

Can I reuse Pierce Protein A/G Magnetic Beads (Cat. No. 88802, 88803)?

Pierce Protein A/G Magnetic Beads are intended for single use only. Therefore, we do not recommend reusing them.

What advantage do Pierce Magnetic Agarose beads have over Dynabeads or Pierce Magnetic Beads?

Pierce Magentic Agarose Beads have a much higher binding capacity.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can I centrifuge the Thermo Scientific Protein A/G Magnetic Beads?

No, centrifuging causes the formation of irreversible aggregates which greatly reduces binding capacity.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How do I reduce non-specific binding to Thermo Scientific Protein A/G Magnetic Beads?

Adding 50-350 mM of NaCl to the Binding/Wash and Elution Buffers can help reduce non-specific bands. Also, use a low-pH elution for single-use applications. The optimal time for low-pH elution is 10 minutes; exceeding 10 minutes may result in non-specific binding and yield reduction.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

When should I avoid boiling the Thermo Scientific Protein A/G Magnetic Beads?

Avoid bead-boil elutions when using rabbit antibodies (primary or secondary) in downstream Western blot applications. Instead, perform elution in SDS-PAGE sample buffer at room temperature. For all other antibody species, boiling the beads in SDS-PAGE sample buffer is acceptable for single-use applications. Boiling could cause bead aggregation and loss of binding activity.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link