Thermo Scientific™

Pierce™ Classic Magnetic IP/Co-IP Kit

Catalog number: 88804
Have Questions?
Thermo Scientific™

Pierce™ Classic Magnetic IP/Co-IP Kit

Catalog number: 88804
Have Questions?
Catalog Number
88804
Unit Size
40 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
8880440 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Classic Magnetic IP/Co-IP Kit enables highly effective and efficient immunoprecipitation (IP) and co-immunoprecipitation (co-IP) of antigens and protein-complexes using less than 10 μg of antibody and a magnetic separator.

Features of the Classic Magnetic IP/Co-IP Kit:

Compatible—magnetic beads based on Protein A/G recombinant protein ensures compatibility with most primary antibodies, whether from mouse or rabbit
Fast—immunoprecipitate in as few as 30 minutes to reduce background and improve the capture of transient protein complexes
Clean—immobilize your antibody to prevent contamination in your eluate
Resistant—no leaching of Protein A/G in the presence of detergents, low pH buffers or common mass spectrometry solvents
Efficient—immunoprecipitate with half the recommended volume of magnetic particles compared to other magnetic beads
Convenient—the comprehensive Co-IP kit contains the magnetic beads and all essential buffers to complete immunoprecipitation experiments

This immunoprecipitation kit uses high-quality Thermo Scientific Pierce Protein A/G Magnetic Beads and optimized buffers and flexible protocol to accomplish high-yield IP or co-IP with manual or automated magnetic-separation tools. The optimized Lysis/Wash Buffer optimizes yield and efficient binding of antibody-antigen and co-IP interactions. The relatively gentle, low-pH Elution Buffer dissociates the bound immune complex from the Protein A/G. Alternatively, the included Lane Marker Sample Buffer provides rapid, denaturing elution for direct SDS-PAGE analysis of IP products.

Includes:
Pierce Protein A/G Magnetic Beads, lysis/wash buffer, elution buffer, neutralization buffer and sample loading buffer

Immunoprecipitation with magnetic particles is performed much like IP with beaded agarose, except that separations are performed using a magnet rather than by centrifugation. The specific antibody is first added to the sample to form an immune complex that is then bound to the magnetic beads. The complex is washed to remove non-bound material and a low-pH elution buffer dissociates the bound immune complex from the Protein A/G. Alternatively, the Lane Marker Sample Buffer is included for dissociation using denaturing conditions or for downstream sample prep for SDS-PAGE analysis. The kit includes Thermo Scientific Pierce Protein A/G Magnetic Beads for fast and convenient magnetic isolation of antigens and optimized buffers for high antigen yield. The beads are removed from the solution manually using a magnetic stand or by automation with an instrument such as the Thermo Scientific KingFisher Flex Instrument.

It is generally known that longer antibody-sample incubations times (2 hours to overnight) usually provide greater yields in IP assays. It is often assumed that this greater yield comes with increased background. Our researchers have found that overnight antibody-sample binding followed by immunoprecipitation with the Pierce Classic Magnetic IP/Co-IP Kit does tend to increase yield but does not increase background. Thus, we recommend that researchers perform this binding step for at least 1 hour whenever possible.

Protocol Summary:
• Incubate cell lysate with IP antibody for 1 to 2 hours at room temperature or overnight at 4ºC.
• Bind antigen-antibody complex to Protein A/G magnetic beads for 1 hour at room temperature.
• Wash beads twice with IP Lysis/Wash Buffer and once with purified water.
• Elute the antigen/antibody complex.

Related Products
Pierce™ Classic IP Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Affinity Purification
For Use With (Equipment)
KingFisher™ Flex, Magnetic Stand
Product Line
Pierce™
Quantity
1 Kit
For Use With (Application)
Immunoprecipitation (IP)
Target
Not Target-Specific
Type
Immunoprecipitation/Co-Immunoprecipitation Kit
Sufficient For
40 Reactions
Format
Kit

Contents & Storage

Sufficient For: 40 IP reactions using 25 μL of beads
• Pierce Protein A/G Magnetic Beads, 1 mL
• Pierce IP Lysis/Wash Buffer, 2 x 50 mL
• Lane Marker Sample Buffer (5X), 5 mL
• Elution Buffer, 5 mL
• Neutralization Buffer, 0.5 mL

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text