Thermo Scientific™

Pierce™ HA-Tag Magnetic IP/Co-IP Kit

Catalog number: 88838
Have Questions?
Thermo Scientific™

Pierce™ HA-Tag Magnetic IP/Co-IP Kit

Catalog number: 88838
Have Questions?
Catalog Number
88838
Unit Size
40 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
8883840 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Magnetic HA-Tag IP/Co-IP Kit contains specific immunoaffinity magnetic beads and reagents to perform immunoprecipitation assays of HA fusion proteins or co-IP experiments using HA-tagged bait proteins.

Features of the HA-Tag Magnetic IP/Co-IP Kit:

Specific magnetic beads—covalently immobilized high-quality anti-HA monoclonal antibody enables high yields of immunoprecipitation products
Low non-specific binding—stable, pre-blocked beads and specific antibody minimize off-target binding
Trouble-free elution—low-pH elution buffer ensures recovery of HA-tagged protein interaction complexes without antibody leaching contamination
Convenient and fast—complete kit and easy-to-follow instructions provide optimized protocols to perform IP or Co-IP experiment in approximately 1 hour
Versatile—magnetic beads are compatible with manual and automated magnetic separation workflows (e.g., Thermo Scientific KingFisher Instruments)

Unlike traditional IP procedures based on capture with Protein A/G beads, this kit uses magnetic beads containing pre-immobilized anti-HA antibody. These Pierce Anti-HA Magnetic Beads ensure specific binding of HA-tagged protein complexes from biological samples. Because the antibody is covalently attached to the beads, IP-targets are easily eluted and recovered without antibody contamination. The complete IP kit includes the magnetic beads, lysis/wash buffer, low-pH elution buffer, neutralization buffer, HA-tag positive control lysate, and non-reducing sample buffer for SDS-PAGE. Protocols are provided for both manual and automated magnetic separation workflows. Sufficient components are provided to perform 40 IP or co-IP assays.

The hemagglutinin (HA) peptide (YPYDVPDYA), derived from the human influenza virus HA protein, is one of several fusion protein tags used for recombinant protein expression. The Pierce Magnetic HA-Tag IP/Co-IP Kit uses a specific, high-affinity immobilized antibody (clone 2-2.2.14) for rapid purification of HA-tagged fusion proteins from bacterial and mammalian cell lysates, as well as from lysates prepared with the Pierce Human in vitro Translation Kits. The beads are incubated with a cell lysate containing HA-tagged protein, the fusion protein is captured, and the beads are subsequently washed and then eluted using low-pH elution buffer or non-reducing sample buffer. The protocol and buffers have been optimized for both IP and co-IP reactions, enriching for specific protein interaction complexes in the eluted samples. Anti-HA antibody can be used to detect HA-tagged protein by Western blot analysis.

For information about the binding capacity and other properties of the magnetic beads used in this kit, see Pierce Anti-HA Magnetic Beads (Part No. 88836).

Related Products
Pierce™ HA-Tag IP/Co-IP Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
Affinity Purification
For Use With (Equipment)
KingFisher™ Flex, Magnetic Stand
Product Line
Pierce™
Quantity
1 Kit
For Use With (Application)
Pull-Down (Tag-Based), Immunoprecipitation (IP)
Target
Target-Specific
Type
Tag IP/Co-IP Kit
Sufficient For
40 Reactions
Format
Kit

Contents & Storage

Sufficient For: 40 IP reactions using 25 μL of anti-HA magnetic beads
• HA-tagged Positive Control (Part No. 26180X), 500 μL
• Pierce Anti-HA Magnetic Beads, 1 mL
• Pierce IP Lysis/Wash Buffer, 2 × 50 mL
• Non-Reducing Sample Buffer (5X), 5 mL
• Elution Buffer (pH 2.0), 5 mL
• Neutralization Buffer (pH 8.5), 1 mL

HA-tagged Positive Control ships separately—store at -70°C. Store remaining components at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text