PT1000 Temperature Probe
PT1000 Temperature Probe
Thermo Scientific™
PT1000 Temperature Probe
For Use With: Thermo Scientific™ Touch Screen Dry Baths/Block Heaters
Have Questions?
Catalog NumberdimVoltage
88870122
Catalog number 88870122
Price (USD)
82.20
Each
-
Add to cart
Price (USD)
82.20
Each
Add to cart
Ask our AI about this Product
Increase lab versatility with the Thermo Scientific™ digital dry baths, which offer a range of configurations with interchangeable modular blocks for a variety of application needs. Designed for basic to advanced applications with a max temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
Thermo Scientific digital dry baths are available in a range of configurations to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment
Applications:
  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification
  • Electrophoresis gel degeneration
  • Serum coagulation
Required Accessory:
  • Unit must be ordered with block(s)
Specifications
DescriptionTemperature Probe, PT1000
For Use With (Equipment)Thermo Scientific™ Touch Screen Dry Baths/Block Heaters
LightingLED
MaterialSteel
Temperature Range (Metric)130°C
TypeThermometer
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link