1-Step Human High-Yield IVT Kit
1-Step Human High-Yield IVT Kit
Thermo Scientific™

1-Step Human High-Yield IVT Kit

The Thermo Scientific 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityType
888902 x 100 μL1-step Human High-yield Mini IVT Kit
8889110 x 100 μL1-step Human High-yield Mini IVT Kit
888922 x 2 mL1-step Human High-yield Maxi IVT Kit
Catalog number 88890
Price (USD)
785.65
Online Exclusive
890.00
Save 104.35 (12%)
Each
-
Add to cart
Quantity:
2 x 100 μL
Type:
1-step Human High-yield Mini IVT Kit
Request bulk or custom format
Price (USD)
785.65
Online Exclusive
890.00
Save 104.35 (12%)
Each
Add to cart
The Thermo Scientific 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greater yield per mL than other mammalian IVT systems.

The 1-Step High-Yield IVT System uses modified HeLa cell extracts to take advantage of the robust human translation machinery and generate functional full-length proteins. In this system, protein expression is performed in a proprietary dialysis device that allows a continuous supply of nucleotides, amino acids and energy generating substrates into the reaction while removing inhibitors of proteins synthesis. This continuous-exchange cell-free (CECF) system enables protein expression in an overnight incubation of up to 750 μg/mL. The complete maxi-scale kit includes all the components required for transcription and translation of a recombinant gene, including an optimized expression vector.

Features of the 1-Step Human High-Yield Maxi IVT Kit:

High expression—up to 750 μg/mL of expressed protein
Reproducibility—low variability between experiments
Fast—express high levels of protein with 6 hours to overnight incubation
Functional—obtain functionally active proteins, including those containing disulfide bonds

1-Step Protein Expression:
Easy-to-use—transcription and translation is performed in one reaction step
Adapt for labeling—amenable to incorporation of heavy or unnatural amino acids

Includes:
• Complete kits contain cell lysate, reaction mix, accessory proteins, 5X dialysis buffer, control DNA, cloning vector, dialysis devices and centrifuge tubes

Requires:
• 30°C incubator or water bath (shaker incubator increases expression by 30 to 50%)

Applications:
• Production of recombinant proteins for structural analysis
• Generate active proteins for protein interaction studies
• Perform rapid expression and characterization of mutant proteins
• Small-scale production of viral particles
• Incorporation of unnatural amino acids or toxic protein production

The 1-Step Human High-Yield IVT Kits are cell-free protein expression systems that provide all of the essential components required for transcription and translation. The kits are optimized with Accessory Proteins and Reaction Mixes that support protein synthesis using a DNA template. The advantages of using the 1-Step Human High-Yield IVT Kits over traditional in vivo expression systems include the ability to express toxic or insoluble proteins, easily perform protein labeling with modified amino-acids and reduce the time and cost of expressing human proteins in tissue culture cells. 1-Step Human IVT Kits are optimized for use with the pT7CFE1 expression vector that utilizes the T7 viral promoter and an EMCV Internal Ribosome Entry Site (IRES) to facilitate high levels of in vitro protein expression. Using a pT7CFE vector is critical for obtaining high expression levels. For added convenience, at family of pT7CFE vectors are available with tandem affinity tags for facilitate protein detection and purification. (For smaller scale and screening options, consider the 1-Step Human Coupled IVT Kit.)

More Product Data
Choosing a vector and purification method for in vitro protein expression
System for high-throughput, high-yield in vitro translation
Expression of highly active proteins using a cell-free human IVT system
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Expressed Protein TypeInsoluble Protein
Quantity2 x 100 μL
Sufficient For2 Reactions of 100 μL Each
Expression SystemCell-Free
Type1-step Human High-yield Mini IVT Kit
Unit SizeEach
Contents & Storage
Sufficient For: 2 reactions of 100 μL each
• HeLa Lysate, 110 μL
• Accessory Proteins, 25 μL
• Reaction Mix, 40 μL
• 5X Dialysis Buffer, 600 μL
• Positive Control DNA (pCFE-GFP), 10 μg
• pT7CFE1-NHis-GST-CHA, 10 μg
• Microdialysis Device, 2 each
• Nuclease-free Water, 5 mL

Store micro-dialysis devices and nuclease-free water at room temperature. Store all other components at -80°C.

Frequently asked questions (FAQs)

When comparing 1-Step Human High-Yield Mini IVT Kit with 1-Step Human Coupled IVT Kit - DNA, is one faster than the other?

No, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA can be run overnight (depends upon the gene).

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Do both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein?

Yes, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I'm using the 1-Step Human High-Yield Mini IVT Kit. Can I get the vector map for the pT7CFE1-NHis-GST-CHA Vector from the kit?

The pT7CFE1-NHis-GST-CHA vector is also offered as a standalone product (Cat. No. 88871) and the vector information can be accessed on the product page (https://www.thermofisher.com/order/catalog/product/88871), under "Vector information" at the bottom of the page.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link