Thermo Scientific™

Pierce™ Glycoprotein Isolation Kit, ConA

Catalog number: 89804
Have Questions?
Thermo Scientific™

Pierce™ Glycoprotein Isolation Kit, ConA

Catalog number: 89804
Have Questions?
Catalog Number
89804
Unit Size
10 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
8980410 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce Glycoprotein Isolation Kit (Concanavilin A) allows isolation of glycoproteins from complex mixtures, including serum, tissue and cultured cell lysates. The procedure uses concanavilin A (ConA) as a ligand to bind a specific class of glycoprotein from complex protein samples—those containing alpha-lined mannose or terminal glucose residues. This kit has been used to isolate glycoproteins from serum as well as HeLa and CHO cell lysates in approximately 50 minutes.

Features of this Glycoprotein Isolation Kit:

High recovery—equivalent or greater glycoprotein recovery versus competitor kits and lectin resins
Fast—glycoprotein purification in less than an hour
Versatile—isolate glycoproteins from various sample types, e.g., human serum and cell lysate.
Robust—lectin does not leach from resin when processing sample
Convenient—complete kit offering lectin resins and spin columns with all necessary reagents
Compatible with Bradford-based protein assays—dialysis or protein precipitation of recovered glycoproteins is not required prior to protein assay

Related Products
Pierce™ Glycoprotein Isolation Kit, WGA
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce Glycoprotein Isolation Kit, ConA
Product Line
Pierce™
Quantity
1 Kit
Target
Glycoprotein
Type
Glycoprotein Isolation Kit
Stationary Phase
Lectin
Sufficient For
10 Reactions
Format
Kit, Microcentrifuge, Spin Column
Column Type
Affinity, Agarose Resin
Unit Size
10 reactions

Contents & Storage

Sufficient For: Isolating glycoproteins with strong affinity for ConA from 10 samples of up to 640 μL (1 to 1.5 mg total protein) each
• ConA Lectin Resin, 1.1 mL
• Binding/Wash Buffer (5X), 6.5 mL
• Elution Buffer, 5 mL
• Spin Columns, 10 columns
• Collection Tubes, 20 tubes

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text