Thermo Scientific™

B-PER™ Complete Bacterial Protein Extraction Reagent

Catalog number: 89822
Have Questions?
Thermo Scientific™

B-PER™ Complete Bacterial Protein Extraction Reagent

Catalog number: 89822
Have Questions?
Catalog Number
89822
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
89822500 mL
-
89821250 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific B-PER Complete Bacterial Protein Extraction Reagent is an easy-to-use cell lysis reagent is a nonionic detergent-based solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation. All B-PER Reagents are compatible with downstream applications, such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay). Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

Features of the B-PER Bacterial Protein Extraction Reagent:

• Detergent in Tris buffer with Lysozyme and Universal Nuclease
• Improved cell lysis and DNA digestion, thereby releasing soluble proteins and reducing viscosity to increase yields
• Recovery of large molecular weight proteins (high MW proteins)
• Cost-effective equivalent to Merck Millipore Bugbuster™ Master Mix

B-PER Complete Bacterial Protein Extraction Reagent is an all-inclusive formulation that combines the lysis reagent with lysozyme and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria. Traditional Tris-buffered and phosphate-buffered formulations without enzymes are also offered. All B-PER Reagents are compatible with downstream applications, such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay). Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

All Thermo Scientific B-PER Protein Extraction Reagents are:
Ready to use—one-step cell lysis of gram-positive and gram-negative bacteria using a mild, nonionic detergent (proprietary) in Tris or phosphate buffer formulations
Fast and simple—just add B-PER Reagent to a bacterial pellet, incubate with mixing for 10 to 15 minutes and recover soluble proteins after pelleting the cell debris
Convenient B-PER Complete Reagent contains lysozyme and a universal nuclease in a single formulation with 4°C storage
Excellent yields—recover recombinant proteins from bacterial lysates or purify inclusion bodies to near-homogeneous levels
Flexible—B-PER Reagents are suitable for any scale of protein extraction and are available in phosphate and 1X and 2X Tris formulations, with and without enzymes
Compatible—completely compatible with addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (e.g., GST, 6xHis) and other applications

Convenient, Ready-to-use Formats
B-PER Bacterial Extraction Reagents are more effective than traditional sonication and typical homemade lysis buffers, many of which include detergents and components that interfere with downstream applications. B-PER Reagents are formulated in Tris or phosphate buffer at physiological pH. They extract native and soluble recombinant proteins and yield lysates that are directly compatible with most downstream workflows such as electrophoresis, affinity purification, immunoprecipitation, protein interaction analysis, crosslinking and protein labeling.

More Product Data
Improved, all-in-one B-PER Reagent for bacterial protein extraction

Related Products
B-PER™ Bacterial Protein Extraction Reagent
B-PER™ Reagent (in Phosphate Buffer)
B-PER™ II Bacterial Protein Extraction Reagent (2X)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
B-PER™
Formulation
Proprietary detergent in 20mM Tris buffer, pH 7.5, containing lysozyme and universal nuclease
Quantity
500 mL
Product Type
Bacterial Protein Extraction Reagent
Volume (Metric)
500 mL

Contents & Storage

Store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text