Pierce™ Protein Transfection Reagent
Pierce™ Protein Transfection Reagent
Thermo Scientific™
Pierce™ Protein Transfection Reagent
Thermo Scientific Pierce Protein Transfection Reagent (formerly Pro-Ject) is a cationic lipid mixture for complexation with proteins, peptides, antibodies andRead more
Catalog NumberQuantity
89850250 μL
Catalog number 89850
Price (JPY)
84,200
Each
Contact Us ›
Quantity:
250 μL
Ask our AI about this Product
Thermo Scientific Pierce Protein Transfection Reagent (formerly Pro-Ject) is a cationic lipid mixture for complexation with proteins, peptides, antibodies and other biologically active molecules to allow their direct intracellular delivery.
Features of Protein Transfection Reagent:
• No need to isolate, clone and transfect gene sequences—Saves days of work
• Optimum delivery in three to four hours after incubation—Fast protein delivery
• Delivery complex is non-covalent—Proteins and peptides retain biological activity after delivery
• Works with proteins, peptides and fluorescently labeled antibodies—Versatility
This is a unique cationic lipid mixture (previously called Pro-Ject Reagent) which, when complexed with proteins or peptides, allows direct intracellular delivery of proteins, peptides, antibodies and other biologically active molecules. Reagent protein complexes attach to negatively charged cell surfaces and either can directly fuse with the membrane and deliver the captured protein into the cell or be endocytosed by the cell and then fuse with the endosome, releasing the captured protein into the cytoplasm. Because the complex formed is noncovalent it does not interfere with the protein's biological activity.
Molecules that have been successfully delivered with the transfection reagent include β-galactosidase, fluorescent-antibody, green fluorescent protein (GFP), high and low molecular weight dextran sulfate, phycoerythrin-BSA, caspase 3, caspase 8 and granzyme B.
Protocol for Optimization of Protein Transfection.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeProtein Transfection Reagent
Product LinePierce™
Quantity250 μL
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C. Product is shipped on dry ice.
Note: For optimal results, do not repeatedly freeze-thaw the β-galactosidase and FITC-antibody controls. Store the control proteins in aliquots suitable for one experiment.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
AE10340151Certificate of AnalysisMay 01, 202589850
AC4611481Certificate of AnalysisApr 15, 202589850
ZH4401331Certificate of AnalysisAug 08, 202489850
ZG4366241Certificate of AnalysisJul 02, 202489850
YK4095431Certificate of AnalysisApr 23, 202489850
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link