Pierce™ Centrifuge Columns
Pierce™ Centrifuge Columns
Thermo Scientific™

Pierce™ Centrifuge Columns

Pack these empty 0.5 to 10 mL spin columns with agarose beads to perform small-scale affinity purification methods in 15- or 50-mL conical microcentrifuge tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
89868800 μL50 Columns
89869B800 μLCustom
89869800 μL4 x 50 Columns
898965 mL25 Columns
898978 mL25 Columns
8989822 mL25 Columns
Catalog number 89868
Price (USD)
101.65
Online Exclusive
116.00
Save 14.35 (12%)
Each
-
Add to cart
Capacity (Metric):
800 μL
Quantity:
50 Columns
Price (USD)
101.65
Online Exclusive
116.00
Save 14.35 (12%)
Each
Add to cart
Ask our AI about this Product

Pierce Centrifuge Columns

Thermo Scientific Pierce centrifuge columns are convenient tools for handling 40 μL to 20 mL of an affinity support for purification. The columns are made from low-protein-binding polypropylene and contain a polyethylene filter (~30-μm pore size) for compatibility with protein purification.

Simply add the affinity resin to the polypropylene column, remove the twist-off bottom, and allow the resin to pack itself. Then add your sample, allow it to bind to the support, and use a centrifuge to efficiently wash away any contaminants and elute your purified protein.

Pierce centrifuge columns can be used in either spin or gravity-flow format. The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and binding steps with dilute samples when slow processing time is desired, column extenders can be used to increase the column reservoir capacity. Use any combination of processing techniques to get the best possible chromatography results.

0.8 mL columns

  • Maximum volume: 800 μL
  • Recommended resin volume: 40–400 μL
  • Receiver tube: fits standard microcentrifuge tubes
  • Cap: O-ring screw-top
  • Twist-off bottom

2 mL columns

  • Maximum volume: 5 mL (2 mL resin bed, 3 mL sample)
  • Recommended resin volume: 400 μL–2 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

5 mL columns

  • Maximum volume: 8 mL (5 mL resin bed, 3 mL sample)
  • Recommended resin volume: 1–5 mL
  • Receiver tube: fits standard 15 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

10 mL columns

  • Maximum volume: 22 mL (10 mL resin bed, 12 mL sample)
  • Recommended resin volume: 2–10 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

20 mL columns

  • Maximum volume: 34 mL (20 mL resin bed, 14 mL sample)
  • Recommended resin volume: 4–20 mL
  • Receiver tube: fits standard 50 mL conical centrifuge tubes
  • Cap: screw-top
  • Twist-off bottom and press-on cap to reseal

Applications

  • Affinity purification, affinity chromatography
  • Immuno-depletion
  • Ion exchange resins
  • Gel filtration, desalting, buffer exchange
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume400 μL
Column TypeEmpty Column, Polyethylene Filter
FormatMicrocentrifuge, Spin Column
Quantity50 Columns
Sample Volume800 μL
Capacity (Metric)800 μL
Product LinePierce™
TypeSpin Column
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link