Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
89880A | 500 mL | 261.65 Online price 266.00 | - |
Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component. Non-specific binding of probe to membrane can be minimized by incubating the membrane in Blocking Buffer for 15 minutes.
Related Products