Tryptic Digestion Kits
Tryptic Digestion Kits
Thermo Scientific™
Tryptic Digestion Kits
Improve peptide digest quality and sensitivity of MS detection using thesw Thermo Scientific Digestion Kits for efficient and reproducible preparation of peptide digests.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
89895Digestion and Guanidination Kit
89871Tryptic Digestion Kit
Catalog number 89895
Price (USD)/ Each
491.65
Online exclusive
506.00 
Save 14.35 (3%)
-
Add to cart
Product Type:
Digestion and Guanidination Kit
Request bulk or custom format
Price (USD)/ Each
491.65
Online exclusive
506.00 
Save 14.35 (3%)
Add to cart
Tryptic Digestion Kits
Catalog number89895
Price (USD)/ Each
491.65
Online exclusive
506.00 
Save 14.35 (3%)
-
Add to cart
Accurate identification and analysis of proteins is a central component of proteomic strategies for studying cellular functions and processes. Using mass spectrometry, proteins and relevant post-translational modifications can be accurately identified, but the method is highly dependent on the quality of the sample preparation. Thermo Scientific Pierce Digestion Kits provide enzymes and reagents for efficient, reproducible digestion and guanidination to improve peptide fragment ionization.
Sample preparation is one of the most variable and time-consuming steps in the analysis of proteins by MS, and the quality and reproducibility of sample extraction and preparation significantly impact the results. Occasionally, additional steps are required to improve sample quality, including digestion and recovery of peptide fragments from SDS-PAGE gels, or peptide guanidination to improve sequence coverage. Each kit has been optimized to produce high quality peptide digests for dependable proteomics analysis using mass spectrometry.
Pierce In-Solution Tryptic Digestion and Guanidination Kit
The Pierce In-Solution Tryptic Digestion and Guanidination Kit provides an optimized procedure and reagents for approximately 90 digests, each containing 0.025 to 10 μg of protein.
Complete digestion—trypsin completely digests protein ranging from 0.025 to 10 μg (∼90 reactions)
Specific—guanidination reaction produces little to no N-terminal modification
Convenient—kit includes reagents for digestion, reduction, alkylation, and guanidination
Efficient—protein can be digested and guanidinated in a single day
Trypsin specifically cleaves peptide bonds at the carboxyl side of arginine and lysine residues. The signals from the arginine-containing peptides are generally stronger, due to the more basic side chain. To enhance overall ionization, guanidination converts lysines to homoarginines and is specific for the amine of lysine, but may occur minimally at the amine of the peptide's N-terminus. This derivatization leads to an increase in the intensity of the lysine-containing peptides and improved sequence coverage overall.
Pierce In-Gel Tryptic Digestion Kit
The Pierce In-Gel Tryptic Digestion Kit provides a complete set of reagents for performing approximately 150 digestions on colloidal Coomassie- or fluorescent dye-stained protein bands. The kit includes modified porcine trypsin, de-staining buffers, reduction reagents, alkylation reagents, and digestion buffers, along with detailed and simple instructions.
Complete—includes all necessary reagents for gel destaining, reduction, alkylation, and tryptic digestion of proteins
Robust—optimized procedure and reagents produce reliable digestions and data generation using a wide range of conditions
Specific—contains highly purified and modified MS-grade trypsin with no chymotryptic activity and minimal autolytic activity
Two key sample preparation steps are the complete digestion of proteins into peptides and subsequent concentration and clean-up of peptides for mass spectrometry analysis. The Pierce In-Gel Tryptic Digestion Kit accomplishes the first of these steps for protein samples extracted from gel slices. Pierce C-18 Spin Columns and Tips accomplish the second step to clean-up the sample to be MS-ready.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Mass Spectrometer
Product TypeDigestion and Guanidination Kit
Shipping ConditionKit is shipped on gel pack.
Starting MaterialProtein Samples, Cell Lysate
Final Product TypePeptide
Detection MethodMass Spectrometry
Workflow StepProtein Digestion
Quantity90 Reactions
Unit SizeEach
Contents & Storage
Upon receipt, store the Pierce Trypsin Protease at -20°C in a nonfrost-free freezer; store other components at 4°C.