1x1 image pixel for data collection
Pierce™ Ubiquitin Enrichment Kit
Pierce™ Ubiquitin Enrichment Kit
Thermo Scientific™

Pierce™ Ubiquitin Enrichment Kit

The Thermo Scientific Pierce Ubiquitin Enrichment Kit facilitates the isolation of polyubiquitin protein conjugates from cultured cells and tissue samples.FeaturesRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
89899Promo Image15 reactions kit
Catalog number 89899
Price (USD)
646.65
Online Exclusive
746.00
Save 99.35 (13%)
Each
In stock
Add to cart
Quantity:
15 reactions kit
Request bulk or custom format
Price (USD)
646.65
Online Exclusive
746.00
Save 99.35 (13%)
Each
Add to cart
The Thermo Scientific Pierce Ubiquitin Enrichment Kit facilitates the isolation of polyubiquitin protein conjugates from cultured cells and tissue samples.

Features of the Ubiquitin Enrichment Kit:

Fast—less than 45 minutes hands-on time for binding and recovery of ubiquitinated proteins
Complete—includes all reagents needed for ubiquitin-modified protein enrichment from cultured cells and tissue samples
Flexible—sample incubation from 2 hours to overnight allows assay to be completed in several hours or in less than 30 minutes after overnight incubation
Robust—compatible with all Thermo Scientific Pierce Cell Lysis Products and standard RIPA formulations
Multiple-sample format—easily processes 1 to 15 samples concurrently

The fractions recovered from the Pierce Ubiquitin Enrichment Kit can be subsequently analyzed to determine the amount of general ubiquitin conjugates present or to identify a specific protein of interest by Western blotting. The assay protocol is fast and straightforward, allowing for isolation of polyubiquitinated proteins and the fractionation of monoubiquitinated species in the resin flow-through. The kit provides exceptional performance as demonstrated by enriching an epoxomicin-treated HeLa lysate for poly ubiquitinated proteins. The Ubiquitin Enrichment Kit outperforms similar resins from other suppliers and provides a clean and specific preparation of proteins when compared to a control resin.

The ubiquitin-proteasome pathway is the principal non-lysosomal pathway that controls the proteolysis of proteins. This pathway is significantly involved in a variety of cellular processes, including DNA repair, transcriptional regulation, signal transduction, cell metabolism and morphogenesis. The ubiquitination process is initiated when ubiquitin, an 8 kDa polypeptide consisting of 76 amino acids, is appended to cellular proteins via the C-terminal glycine of the ubiquitin molecule. Following an initial monoubiquitination event, the formation of a ubiquitin polymer may occur. Polyubiquitinated proteins are then recognized by the 26S proteasome that catalyzes the degradation of the ubiquitinated protein and the recycling of ubiquitin. Differences in total ubiquitination or the ubiquitination of specific proteins affect numerous pathological conditions, including malignancies, certain genetic diseases and neurodegenerative diseases.¹
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeAffinity, Agarose Resin
FormatKit, Microcentrifuge, Spin Column
Quantity15 reactions kit
Stationary PhaseProprietary Ligand
Sufficient For15 Reactions
TargetPolyubiquitinated Protein
Product LinePierce™
TypeUbiquitin Enrichment Kit
Unit SizeEach
Contents & Storage
Sufficient For: Enriching polyubiquitinated proteins from 15 lysate samples, each containing 0.15 mg total protein
• Polyubiquitin Positive Control (1000X), 50 μL (store at -20°C)
• Anti-ubiquitin Antibody (rabbit antiserum), 5 μL (store at -20°C)
• Polyubiquitin Affinity Resin Slurry, 300 μL (store at 4°C—do not freeze)
• TBS Pack (makes 500mL), 1 pack (store at 4°C)
• Spin Columns and Accessories, 18 columns (store at 4°C)

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link