RIPA Lysis and Extraction Buffer
New Protein Purification and Isolation Support Center—Find technical tips and troubleshooting recommendations from our scientists.
RIPA Lysis and Extraction Buffer
Thermo Scientific™

RIPA Lysis and Extraction Buffer

Thermo Scientific RIPA Lysis and Extraction Buffer is a high-quality, ready-to-use and fully disclosed formulation of a popular cell lysisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
89900100 mL
89901250 mL
Catalog number 89900
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific RIPA Lysis and Extraction Buffer is a high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent for cultured mammalian cells.

Features of RIPA Buffer:

Convenient—ready-to-use solution; no need to assemble and prepared components yourself
Flexible—compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification
Versatile—enables extraction of cytoplasmic, membrane and nuclear proteins
Disclosed formulation—contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues

This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells. The popular reagent enables the extraction of membrane, nuclear and cytoplasmic proteins and is compatible with many applications, including reporter assays, the Thermo Scientific BCA Protein Assay, immunoassays and protein purification. Inhibitors such as Thermo Scientific Halt Protease Inhibitor Cocktail (Part No. 78430) and Halt Phosphatase Inhibitor Cocktail (Part No. 78420) are also compatible with this RIPA buffer formulation and can be added before use to prevent proteolysis and maintain protein phosphorylation.

RIPA buffer derives its name from the original application for which it was developed: the radio-immunoprecipitation assay. While this isotopic assay method is rarely performed in laboratories today, the acronym for this lysis buffer formulation has endured in common use. RIPA cell lysis reagent is highly effective for protein extraction from a variety of cell types because it contains three non-ionic and ionic detergents. One disadvantage of this detergent formulation is its relative incompatibility with certain downstream applications compared to other lysis reagents.

Related Products
Pierce™ IP Lysis Buffer
M-PER™ Mammalian Protein Extraction Reagent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLiquid
Quantity100 mL
Volume (Metric)100 mL
Product TypeExtraction Buffer
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link