You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Protein Analysis Reagents
Antibody Production and Purification
Special Purpose IgG Purification Resins and Kits
Melon™ Gel Regenerant
Thermo Scientific™
Melon™ Gel Regenerant
Catalog number:
89973
Have Questions?
Contact Us
Thermo Scientific™
Melon™ Gel Regenerant
Catalog number:
89973
Have Questions?
Contact Us
Catalog Number
89973
Unit Size
1 pack
Price (USD)
46.91
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
89973
1 pack
46.91
-
Save to list
Add to cart
Request bulk or custom format
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Melon Gel Regenerant, formulation: proprietary powder dry-blend,sufficient for preparing 500 mL of regenerant.
Related Products
Melon™ Gel IgG Spin Purification Kit
Melon™ Gel IgG Purification Kit
Melon™ Gel Purification Buffer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
Gel Regenerant
Product Line
Melon™
Reagent Type
Regeneration Buffer
Description
Melon Gel Regenerant
Quantity
1 Pack
Form
Powder
Contents & Storage
2°C to 8°C
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search