Informational:Note: This product will be discontinued soon. Its replacement, catalog #88364 (DMEM for SILAC) is available for online purchase now.
Thermo Scientific™
DMEM Media for SILAC
Catalog number: 89985
Have Questions?
Thermo Scientific™
DMEM Media for SILAC
Catalog number: 89985
Have Questions?
Catalog Number
89985
Unit Size
Case of 30
Price (USD)
Catalog NumberUnit SizePrice (USD)
89985Case of 30
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific DMEM Media for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
Features of DMEM Media for SILAC :
Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
High quality—media are sterile, endotoxin-free and cell culture-compatible
Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of "heavy" 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.
This SILAC media is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Contents & Storage
Store in refrigerator (2 to 8°C).