Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
900-K68K consists of: 900-K68 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
Mouse Eotaxin ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant mouse Eotaxin in a sandwich ELISA format. Mouse Eotaxin ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Ccl11, eotaxin, Scya11
Gene ID : (Mouse) 20292
Gene symbol : Ccl11
Protein Aliases : C-C motif chemokine 11, Eosinophil chemotactic protein, Eotaxin, Eotaxin 1, Eotaxin1, MGC22554, small chemokine (C-C motif) ligand 11, small inducible cytokine A11, Small-inducible cytokine A11
UniProt ID (Mouse) P48298
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support