Orion™ pH Electrode Cleaning Solutions
Orion™ pH Electrode Cleaning Solutions
Actual product may vary
Thermo Scientific™

Orion™ pH Electrode Cleaning Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFilling Solution TypeQuantity
900022-WABacteria Removal (B)4 x 60 mL
510015No Cal Stain Lifter60 mL
900020A, B, C, D60 mL (B, D), 30 mL (A, C)
900021-WAProtein Removal (A)4 x 30 mL
900023General Cleaning (C)4 x 30 mL
900024Oil and Grease Removal (D)4 x 60 mL
911110Rinse10 pk.
911125Rinse25 pk.
Catalog number 900022-WA
Price (EUR)
-
Filling Solution Type:
Bacteria Removal (B)
Quantity:
4 x 60 mL

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.

Cleaning Solutions:

 

  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for removing bacterial contaminants, cleaning solution C is for general cleaning and cleaning solution D is for removing oil and grease contaminants
  • Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution

Includes (PN 900020):

  • One bottle each of cleaning solutions A, B, C and D
  • One beaker
  • One pipet

Note:

The following part numbers have been changed due to a recent system upgrade:

Old Part Number: 900021

New Part Number: 900021-WA

Old Part Number: 900022

New Part Number: 900022-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.

Warranty: Out of box


Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description4 x 2 oz. (4 x 60mL) Bottles of Cleaning Solution B for Bacteria Removal, Beaker and Pipet
Filling Solution TypeBacteria Removal (B)
For Use With (Application)Solutions restore proper electrode performance and prolong the useful life of the electrode
PackagingBotlle
Quantity4 x 60 mL
For Use With (Equipment)pH Electrode
TypeCleaning Solution
Unit SizeCase of 4

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link