Thermo Scientific™

MALDI Matrix Sample Pack, Single-Use

Catalog number: 90035
Have Questions?
Thermo Scientific™

MALDI Matrix Sample Pack, Single-Use

Catalog number: 90035
Have Questions?
Catalog Number
90035
Unit Size
24 tubes
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
9003524 tubes
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization. This MALDI Matrix Sample Pack contains 8 tubes each of CHCA, SA and DHB reagents.

Features of MALDI Matrices:

• Single-use packaging saves time and reduces waste
• Provide accurate quantity of matrix every time

Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The unique packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes it easy to prepare high-quality MALDI reagents in minutes.

MALDI and electrospray ionization (ESI) are the most common methods to ionize peptides for MS analysis. MALDI is widely used because it is significantly faster than ESI and the ionized molecules are almost exclusively singly charged, making interpretation easier. Also, MALDI is readily automated and highly sensitive. This powerful method for MS peptide analysis is now easier to perform with our single-use, highly pure MALDI matrices that are exceptionally convenient to use.

Related Products
CHCA MALDI Matrix, Single-Use
SA MALDI Matrix, Single-Use
DHB MALDI Matrix, Single-Use
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce MALDI Matrix Sample Pack
For Use With (Equipment)
Mass Spectrometer
Product Type
MALDI Matrix
Starting Material
Small Molecules
Final Product Type
Small Molecules
Detection Method
Mass Spectrometry
Workflow Step
Calibration

Contents & Storage

Upon receipt store product at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text