Applied Biosystems™

GeneChip™ Hybridization Control Kit

Catalog number: 900454
Have Questions?
Applied Biosystems™

GeneChip™ Hybridization Control Kit

Catalog number: 900454
Have Questions?
Catalog Number
900454
Unit Size
1 kit
Price (USD)
Catalog NumberUnit SizePrice (USD)
9004541 kit
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneChip™ Hybridization Control Kit is required for use with GeneChip eukaryotic expression arrays

Each kit contains 20X Hybridization Control Stock composed of pre-mixed biotin-labeled bioB, bioC, bioD and cre, in staggered concentrations, ready to be added directly to the hybridization cocktail. These controls facilitate monitoring of the hybridization process for troubleshooting.

In addition, this kit also contains Control Oligo B2, used to provide alignment signals for image analysis. (Additional Oligo B2 can be ordered separately.)

All kit components have been thoroughly tested to ensure they meet our high-quality standards.

Labeled with the GeneChip IVT Labeling Kit, these control kits replace the previous generation Hybridization Control Kits.

The GeneChip™ Hybridization Control Kit is also available as part of the Complete Reagent Packages for Eukaryotic Arrays.

Related Links
B2 Control Oligo (3nM)
GeneTitan™ Hybridization, Wash, and Stain Kit for WT Array Plates
GeneTitan™ Hybridization, Wash, and Stain Kit for 3' IVT Arrays
GeneChip™ miRNA 4.1 24-Array Plate and Trays
GeneChip™ miRNA 4.1 96-Array Plate and Trays
GeneChip™ miRNA 3.1 24-Array Plate and Trays
GeneChip™ miRNA 3.1 96-Array Plate and Trays
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Hybridization Control Kit
Format
Array Cartridge
For Use With (Application)
Transcriptome Profiling
Product Line
GeneChip™
Reagent Type
Hybridization Control

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text