Applied Biosystems™
GeneChip™ Hybridization Control Kit
Catalog number: 900457
Have Questions?
Applied Biosystems™
GeneChip™ Hybridization Control Kit
Catalog number: 900457
Have Questions?
Catalog Number
900457
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
9004571 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneChip™ Hybridization Control Kit is required for use with GeneChip eukaryotic expression arrays
Each kit contains 20X Hybridization Control Stock composed of pre-mixed biotin-labeled bioB, bioC, bioD and cre, in staggered concentrations, ready to be added directly to the hybridization cocktail. These controls facilitate monitoring of the hybridization process for troubleshooting.
In addition, this kit also contains Control Oligo B2, used to provide alignment signals for image analysis. (Additional Oligo B2 can be ordered separately.)
All kit components have been thoroughly tested to ensure they meet our high-quality standards.
Labeled with the GeneChip IVT Labeling Kit, these control kits replace the previous generation Hybridization Control Kits.
The GeneChip™ Hybridization Control Kit is also available as part of the Complete Reagent Packages for Eukaryotic Arrays.
Related Links
B2 Control Oligo (3nM)
GeneTitan™ Hybridization, Wash, and Stain Kit for WT Array Plates
GeneTitan™ Hybridization, Wash, and Stain Kit for 3' IVT Arrays
GeneChip™ miRNA 4.1 24-Array Plate and Trays
GeneChip™ miRNA 4.1 96-Array Plate and Trays
GeneChip™ miRNA 3.1 24-Array Plate and Trays
GeneChip™ miRNA 3.1 96-Array Plate and Trays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
Oligonucleotides, Primers
Probe Classification
Eukaryotic Expression Probe
Product Line
GeneChip™