GeneChip™ Zebrafish Genome Array
GeneChip™ Zebrafish Genome Array
Applied Biosystems™
GeneChip™ Zebrafish Genome Array
The GeneChip™ Zebrafish Genome Array can be used to study gene expression of over 14,900 Danio rerio transcripts. Sequence informationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90048830 arrays
9004875 arrays
Catalog number 900488
Price (USD)
-
Number of Arrays:
30 arrays
The GeneChip™ Zebrafish Genome Array can be used to study gene expression of over 14,900 Danio rerio transcripts. Sequence information for this array was selected from the following public data sources: RefSeq (July 2003), GenBank™ (release 36.0, June 2003), dbEST (July 2003), and UniGene (Build 54, June 2003). Probe sets on the array were designed with 6 oligonucleotide pairs to detect each transcript.
This array was designed in collaboration with representative members of the Zebrafish community and the National Institutes of Health.
Note: The DsRed probe set is provided with permission from BD Biosciences, and BD Biosciences grants users a limited license to utilize this probe set only on this array. Other uses of the probe set, or other DsRed sequence or sequences requires a license from BD Biosciences.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip™
Quantity30 arrays
TypeZebrafish Genome Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays30 arrays
SpeciesZebrafish
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection