Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Asp-N Endoproteinase, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Protease |
Product Line | Pierce™ |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 2 μg |
Unit Size | 2 µg |
Description | Chymotrypsin Endoproteinase, MS Grade (TLCK-treated) |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Protease |
Product Line | Pierce™ |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 4 x 25 μg |
Unit Size | 4 x 25 µg |
Description | Glu-C Endoproteinase, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Protease |
Product Line | Pierce™ |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 5 x 10 μg |
Unit Size | 5 x 10 µg |
Description | Trypsin Protease, MS Grade (Frozen) |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Trypsin Protease |
Product Line | Pierce™ |
Shipping Condition | Products are shipped with ice. |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Protease (Enzyme) |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 1 x 100 μg |
Form | Liquid |
Unit Size | 1 x 100 µg |
Description | Trypsin Protease, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Trypsin Protease |
Product Line | Pierce™ |
Shipping Condition | Products are shipped with ice. |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Enzyme |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 1 mg |
Form | Powder |
Unit Size | 1 mg |
Description | Trypsin/Lys-C Protease Mix, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Trypsin/Lys-C Protease Mix |
Product Line | Pierce™ |
Shipping Condition | Wet Ice |
Starting Material | Protein Samples |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 20 μg |
Form | Solid |
Unit Size | 20 µg |
Description | Trypsin/Lys-C Protease Mix, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Trypsin/Lys-C Protease Mix |
Product Line | Pierce™ |
Shipping Condition | Wet Ice |
Starting Material | Protein Samples |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 5 x 20 μg |
Form | Solid |
Unit Size | 5 x 20 µg |
Description | Trypsin/Lys-C Protease Mix, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Trypsin/Lys-C Protease Mix |
Product Line | Pierce™ |
Shipping Condition | Wet Ice |
Starting Material | Protein Samples |
Final Product Type | Peptides |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 100 μg |
Form | Solid |
Unit Size | 100 µg |
Description | Lys-C Endoproteinase, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Protease |
Product Line | Pierce™ |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 1 x 20 μg |
Unit Size | 20 µg |
Description | Lys-C Protease, MS Grade |
For Use With (Equipment) | Mass Spectrometer |
Product Type | Protease |
Product Line | Pierce™ |
Shipping Condition | Wet Ice |
Starting Material | Protein Samples, Cell Lysate |
Final Product Type | Peptide |
Detection Method | Mass Spectrometry |
Workflow Step | Protein Digestion |
Quantity | 100 μg |
Unit Size | 100 µg |
Catalog Number | Specifications | Unit Size | Description | Quantity | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
90053 | 2 µg | Asp-N Endoproteinase, MS Grade | 2 μg | 304.65 Online price 317.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90056 | 4 x 25 µg | Chymotrypsin Endoproteinase, MS Grade (TLCK-treated) | 4 x 25 μg | 285.65 Online price 303.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90054 | 5 x 10 µg | Glu-C Endoproteinase, MS Grade | 5 x 10 μg | 276.65 Online price 294.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90305 | 1 x 100 µg | Trypsin Protease, MS Grade (Frozen) | 1 x 100 μg | 68.65 Online price 72.25 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90059 | 1 mg | Trypsin Protease, MS Grade | 1 mg | 710.65 Online price 756.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A40007 | 20 µg | Trypsin/Lys-C Protease Mix, MS Grade | 20 μg | 69.65 Online price 73.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A41007 | 5 x 20 µg | Trypsin/Lys-C Protease Mix, MS Grade | 5 x 20 μg | 194.65 Online price 206.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A40009 | 100 µg | Trypsin/Lys-C Protease Mix, MS Grade | 100 μg | 179.65 Online price 191.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90051 | 20 µg | Lys-C Endoproteinase, MS Grade | 1 x 20 μg | 350.65 Online price 373.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90307 | 100 µg | Lys-C Protease, MS Grade | 100 μg | 1,218.65 Online price 1,296.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|