GeneChip™ Human Exon 1.0 ST Array
GeneChip™ Human Exon 1.0 ST Array
Applied Biosystems™
GeneChip™ Human Exon 1.0 ST Array
Explore GeneChip-compatible Software for Exon-level AnalysisWith approximately four probes per exon and roughly 40 probes per gene, the GeneChip™ HumanMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoNúmero de arrays
90065130 arrays
9006506 arrays
Número de catálogo 900651
Precio (EUR)
30.420,00
Each
-
Añadir al carro de la compra
Número de arrays:
30 arrays
Precio (EUR)
30.420,00
Each
Añadir al carro de la compra
Ask our AI about this Product
Explore GeneChip-compatible Software for Exon-level Analysis
With approximately four probes per exon and roughly 40 probes per gene, the GeneChip™ Human Exon 1.0 ST Array enables two complementary levels of analysis—gene expression and alternative splicing. Multiple probes per exon enable 'exon-level' analysis and allow you to distinguish between different isoforms of a gene. This exon-level analysis on a whole-genome scale opens the door to detecting specific alterations in exon usage that may play a central role in disease mechanism and etiology. The second level is 'gene-level' expression analysis, in which multiple probes on different exons are summarized into an expression value of all transcripts from the same gene. Exon arrays provide the most comprehensive coverage of the genome, including empirically supported and predicted transcribed sequences, enabling the discovery of previously unidentified novel events.
Features of this integrated solution include:
• Highest resolution expression analysis, interrogating over 1 million exon clusters within the known and predicted transcribed regions of the entire genome. Read more
• Tailored GeneChip Whole Transcript (WT) Assay and Reagents utilizing a random priming method for generating sense targets along the entire length of the transcripts.
• Flexible data analysis solutions and comprehensive annotation, giving you the opportunity to explore and dissect the data in various user-defined workflows.
Learn More
Explore the Exon Array Design
GeneChip Exon Array Sample Data
Technical Note, Array Design for the Human Exon 1.0 ST Array (pdf, 348 KB)
Technical Note, Identifying and Validating Alternative Splicing Events
XRAY demos for exon expression analysis
View a two-minute demo on a typical workflow using Biotique's XRAY for the analysis of GeneChip Exon Arrays
Windows Media Player | Quicktime
View a two-minute demo on the interpretation of GeneChip; Exon Array data from Biotique's XRAY
Windows Media Player | Quicktime
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
TipoHuman Exon 1.0 ST Array
matriz, red, conjuntoTranscriptome Profiling
Número de arrays30 arrays
FormatoArray Cartridge
EspecieHumano
Línea de productosGeneChip™
Cantidad30 arrays
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documentos y descargas
Certificados
Hojas de datos de seguridad
Compartir número de catálogo, nombre o enlace.