Applied Biosystems™

GeneChip™ WT Terminal Labeling Kit

Catalog number: 900670
Have Questions?
Applied Biosystems™

GeneChip™ WT Terminal Labeling Kit

Catalog number: 900670
Have Questions?
Catalog Number
900670
Unit Size
10 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
30 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
90067010 reactions
-
90067130 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
GeneChip™ WT Sense Target Labeling and Control Reagents (900652) and GeneChip WT cDNA Synthesis and Amplification Kits (900672 and 900673) are being discontinued in June 2010.

The Whole Transcript (WT) Sense Target Labeling Assay has an updated protocol. The GeneChip WT (Whole Transcript) Terminal Labeling Kit is optimized to be used for the fragmentation and labeling steps of the GeneChip WT Sense Target (ST) Labeling Assay.

The WT Terminal Labeling Kit employs a strategy for reproducible DNA fragmentation with a combination of uracil DNA glycosylase (UDG) and apurinic/apyrimidinic endonuclease 1 (APE 1). The typical input material is random-primed, single-stranded DNA

Following fragmentation, the resulting fragmented DNA can then be labeled using terminal deoxynucleotidyl transferase (TdT) in the presence of a proprietary biotinylated compound, GeneChip DNA Labeling Reagent. Both components are also provided in the WT Terminal Labeling Kit. The final labeled DNA target is ready to be added to the cocktail for hybridizing to the arrays such as the GeneChip Human Exon 1.0 ST Array.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Biotin-based
Format
Kit
Final Product Type
DNA (Fragmented, Labeled)
Labeling Target
Random-primed DNA (Single-stranded)
Includes Label or Dye
Direct Labeling
No. of Reactions
10
Product Line
GeneChip™
Product Type
WT Terminal Labeling Kit

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text