Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Thermo Scientific™
Pierce™ S-Nitrosylation Western Blot Kit and HENS Buffer
Thermo Scientific Pierce S-Nitrosylation Western Blot Kit enables sensitive detection of protein S-nitrosocysteine post-translational modifications in a complete, easy-to-use kit or stand-alone HENS buffer for S-Nitrosylation Western Blot Kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
9010540 ReactionsWestern Blot Kit
90106100 mLHENS Buffer
Catalog number 90105
Price (USD)
476.00
Special offer
Online exclusive
Ends: 15-Aug-2025
560.00
Save 84.00 (15%)
Each
In stock
Add to cart
Quantity:
40 Reactions
Product Type:
Western Blot Kit
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
476.00
Special offer
Online exclusive
Ends: 15-Aug-2025
560.00
Save 84.00 (15%)
Each
Add to cart
Ask our AI about this Product
The Pierce S-Nitrosylation Western Blot Kit provides cell lysis/reaction buffer, sulfhydryl-reactive blocking agent, reducing agent, labeling agent, and anti-TMT antibody to label 40 samples (100 μg each). As with traditional S-nitrosylation switch assays, unmodified cysteines are blocked by a sulfhydryl-reactive compound. S-nitrosylated cysteines are selectively reduced with ascorbate in HENS Buffer for labeling with iodoTMTzero reagents, which irreversibly bind to the cysteine thiol that was S-nitrosylated. Detection of the TMT reagent-modified proteins is facilitated using an anti-TMT antibody.
S-nitrosylation is a post-translational modification that regulates numerous processes, including cellular proliferation, apoptosis, smooth muscle relaxation, neurotransmitter release, and differentiation. During S-nitrosylation, nitric oxide radicals react with cysteine thiols to produce an S-NO adduct that alters protein activity.
Pierce S-Nitrosylation Western Blot Kit
The Pierce S-Nitrosylation Western Blot Kit provides the necessary reagents to perform a modified version of the traditional S-nitrosylation switch assay, including a cysteine-blocking reagent, reducing agent, reaction buffer, iodoTMT labeling reagent, and anti-TMT antibody. Labeling is accomplished using the non-biological iodoTMT reagent instead of HPDP-biotin, reducing background signal during western blot detection. In addition to detection of S-nitrosylated proteins by western blot, an immobilized anti-TMT resin can be used to selectively enrich S-nitrosylated proteins/peptides labeled with iodoTMT reagents. This workflow allows for S-nitrosylation site mapping and multiplexed quantitation using mass spectrometry.
Features of Pierce S-Nitrosylation Western Blot Kit include:
Specific—optimized labeling reactions switch S-nitrosocysteine post translational modifications with iodoTMT reagent for easy detection by western blot using anti-TMT antibody
Better signal-to-noise—uses a non-biological iodoTMT reagent for labeling instead of HPDP-biotin, resulting in less background during western blot detection
Workflow-compatible—labeling method is compatible with alternative methods of analysis, including enrichment via anti-TMT resin and multiplex quantitation by mass spectrometry
Convenient—all needed reagents included one kit
HENS Buffer
HENS Buffer consists of 100 mM HEPES (pH 7.8), 1 mM EDTA, 0.1 mM Neocuproine, and 1% SDS. It is a cell lysis buffer intended for use with the Pierce S-Nitrosylation Western Blot Kit, which enables sensitive detection of protein S-nitrosocysteine post-translational modifications in a complete, easy-to-use kit.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ReactivityAll species
Detection MethodS-Nitrosylation
Product LinePierce™
Product TypeWestern Blot Kit
Quantity40 Reactions
Unit SizeEach
Contents & Storage
Upon receipt store kit at –20°C, or store individual components as indicated above. Kit is shipped with an ice pack.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
AB396267Certificate of AnalysisFeb 28, 202590106
ZL393742Certificate of AnalysisJan 31, 202590105
ZD389393Certificate of AnalysisApr 25, 202490106
ZC386480Certificate of AnalysisApr 12, 202490105
ZA386431Certificate of AnalysisFeb 14, 202490106
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection