Applied Biosystems™

GeneChip™ Human Gene 1.0 ST Array

Catalog number: 901087
Have Questions?
Applied Biosystems™

GeneChip™ Human Gene 1.0 ST Array

Catalog number: 901087
Have Questions?
Catalog Number
901087
Unit Size
30 arrays
Price (USD)
Catalog Number
Unit Size
2 arrays
Price (USD)
Catalog Number
Unit Size
6 arrays
Price (USD)
Catalog NumberUnit SizePrice (USD)
90108730 arrays
9010852 arrays
9010866 arrays
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneChip™ Human Gene 1.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and long intergenic non-coding RNA transcripts. We also offer the GeneChip Human Gene 2.0 ST Array, which can measure differential expression of long intergenic non-coding RNA transcripts.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Human Gene 1.0 ST Array
Array
Transcriptome Profiling
Number of Arrays
30 arrays
Format
Array Cartridge
Species
Human
Product Line
GeneChip™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text