1x1 image pixel for data collection
Capitol Vial Single Specimen Urine Collection Kit
Capitol Vial Single Specimen Urine Collection Kit
Capitol Vial Single Specimen Urine Collection Kit
Capitol Vial Single Specimen Urine Collection Kit
Capitol Vial Single Specimen Urine Collection Kit
Capitol Vial Single Specimen Urine Collection Kit
Capitol Vial Single Specimen Urine Collection Kit
Thermo Scientific™
Capitol Vial Single Specimen Urine Collection Kit
Collect, store and transport urine samples for drugs of abuse testing with Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kits.
Have Questions?
Catalog NumberSterility
90111TSSterile
Catalog number 90111TS
Price (USD)
158.40
Online Exclusive
189.00
Save 30.60 (16%)
Case of 100
Estimated availability date  Pending  
Add to cart
Sterility:
Sterile
Price (USD)
158.40
Online Exclusive
189.00
Save 30.60 (16%)
Case of 100
Add to cart
Thermo Scientific™ Capitol Vial Single Specimen Urine Collection Kits provide a collection beaker, transport vial, and specimen transport bag in a tamper evident package to support drug testing programs. The kits are constructed from a low-binding polypropropylene resin that has been laboratory tested for interaction with drugs of abuse analytes.
  • Easy-to-use—simply peel away the foil-top lid, remove the components and provide donor with the collection cup
  • After specimen is collected, transfer urine into the tamper-evident vial, close the hinged lid until it snaps, and bag for transport
Ordering Information:
Case of 100. Custom branding available for foil tops and specimen bags.
Includes:
Collection cup (8 oz.) with temperature indicator strip, 45mL Tamper evident transport vial, dual pocket specimen bag with absorbent pad, and a foil top seal
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (Metric)45 mL
DescriptionSingle Specimen Collection Kit
IncludesCollection cup (8 oz.) with temperature indicator strip, 45 mL Tamper evident transport vial, Generic dual pocket specimen bag, Absorbent pad and Foil top
MaterialPolypropylene
Product TypeSpecimen Collection Kit
SterilitySterile
Unit SizeCase of 100
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link