TMT10plex™ Isobaric Label Reagents and Kits
TMT10plex™ Isobaric Label Reagents and Kits
TMT10plex™ Isobaric Label Reagents and Kits
TMT10plex™ Isobaric Label Reagents and Kits
TMT10plex™ Isobaric Label Reagents and Kits
TMT10plex™ Isobaric Label Reagents and Kits
Thermo Scientific™

TMT10plex™ Isobaric Label Reagents and Kits

TMT10plex Isobaric Label Reagents and Kits contain ten isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm, and a mass reporter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
901133 x 0.8 mgIsobaric Mass Tag Labeling Kit
901101 x 0.8 mg (per tag), 10 reactionsIsobaric Label Reagent Set
901113 x 0.8 mg (per tag), 30 reactionsIsobaric Label Reagent Set
904061 x 5 mg (per tag), 60 reactionsIsobaric Label Reagent Set
903098 x 0.2 mgIsobaric Label Reagent Set
Catalog number 90113
Price (USD)
3,340.50
Special offer
Online exclusive
Ends: 15-Aug-2025
3,930.00
Save 589.50 (15%)
Each
Add to cart
Quantity:
3 x 0.8 mg
Product Type:
Isobaric Mass Tag Labeling Kit
Request bulk or custom format
Price (USD)
3,340.50
Special offer
Online exclusive
Ends: 15-Aug-2025
3,930.00
Save 589.50 (15%)
Each
Add to cart
TMT10plex Isobaric Label Reagents and Kits contain ten isobaric compounds with the same mass and chemical structure (i.e., isotopomeric) composed of an amine-reactive NHS-ester group, a spacer arm, and a mass reporter. Up to ten different peptide samples prepared from cells or tissues can be labeled in parallel and then combined for analysis. For each sample, a unique reporter mass (i.e., TMT10 126-131 Da) in the low-mass region of the high-resolution MS/MS spectrum is used to measure relative protein expression levels during peptide fragmentation and tandem mass spectrometry.

The Tandem Mass Tag (TMT) Isobaric Label reagents and kits are specifically designed to enable identification and quantitation of proteins in different samples using tandem mass spectrometry. The TMT10plex label reagents share an identical structure with TMTzero, TMTduplex, and TMTsixplex reagents, but contain different numbers and combinations of 13C and 15N isotopes in the mass reporter. The different isotopes result in a 10-plex set of tags that have mass differences in the reporter that can be detected using high resolution Orbitrap MS instruments.

Advantages of the TMT10plex label reagents include increased multiplex relative quantitation, increased sample throughput, and fewer missing quantitative values among samples. TMT10plex label reagents are ideal for analysis of multiple protein samples from inhibitor dose response experiments, time course experiments, or improving statistics on biological replicates.

Features of TMT10plex Isobaric Label reagent sets and mass tagging kits
Powerful—multiplexed MS analysis increases sample throughput and enables relative quantitation of up to ten different samples derived from cells, tissues, or biological fluids
Consistent—identical reagent structure and performance among TMTzero, TMTduplex, TMTsixplex, and TMT10plex reagents allows efficient transition from method development to multiplex quantitation, enabling biomarker discovery research across platforms and datasets
Robust—increased multiplex capability results in fewer missing quantitative values among samples and higher confidence among replicates
Efficient—amine-reactive, NHS-ester-activated reagents ensure efficient labeling of all peptides regardless of protein sequence or proteolytic enzyme specificity
Compatible—optimized for use with high resolution Orbitrap MS/MS platforms, with data analysis fully supported by Proteome Discoverer 2.3.

Applications:
• Protein identification and quantitation from multiple samples of cells, tissue, or biological fluids
• Protein expression profiling of normal vs. disease states or control vs. treated
• Quantitative analysis of proteins for which no antibodies are available
• Identification and quantitation of membrane and post-translationally modified proteins
• Identification and quantification of hundreds to thousands of proteins in a single experiment

TMT Reagents are provided as standalone sets, a 96-well format, or in optimized kit formats containing all necessary reagents and controls for maximum flexibility, convenience, and reliability. When combined with the industry-leading, high resolution Thermo Scientific Orbitrap instruments and software, TMT10plex reagents provide integrated total solutions for quantitative protein expression analysis.

For Research Use Only. Not for use in diagnostic procedures.

General References:

  1. Thompson, A., et al.(2003). Tandem mass tags: a novel quantification strategy for comparative analysis of complex protein mixtures by MS/MS. Anal. Chem.75(8), 1895-1904.
  2. Dayon, L., et al.(2008). Relative quantification of proteins in human cerebrospinal fluids by MS/MS using 6-plex isobaric tags. Anal. Chem. 80(8), 2921 -2931.
Specifications
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeTandem Mass Tag™ (TMT) Labels
Quantity3 x 0.8 mg
Sufficient For30 Reactions
Workflow StepProtein Labeling
Detection MethodMass Spectrometry
Product LineTMT10plex™
Product TypeIsobaric Mass Tag Labeling Kit
Starting MaterialCell Lysate
Unit SizeEach
Contents & Storage
Sufficient for: Three 10-plex (3 x 10-way) experiments
• TMT10-126 Label Reagent, 3 x 0.8 mg
• TMT10-127N Label Reagent, 3 x 0.8 mg
• TMT10-127C Label Reagent, 3 x 0.8 mg
• TMT10-128N Label Reagent, 3 x 0.8 mg
• TMT10-128C Label Reagent, 3 x 0.8 mg
• TMT10-129N Label Reagent, 3 x 0.8 mg
• TMT10-129C Label Reagent, 3 x 0.8 mg
• TMT10-130N Label Reagent, 3 x 0.8 mg
• TMT10-130C Label Reagent, 3 x 0.8 mg
• TMT10-131 Label Reagent, 3 x 0.8 mg
• Dissolution Buffer (1M triethyl ammonium bicarbonate), 5 mL
• Denaturing Reagent (10% SDS), 1 mL
• Reducing Reagent (0.5M TCEP), 1 mL
• Iodoacetamide, 12 x 9 mg
• Quenching Reagent (50% hydroxylamine), 1 mL
• Pierce Trypsin Protease, MS Grade, 5 x 20 μg
• Trypsin Storage Solution, 250 μL
• Albumin, Bovine, 2.5 mg

Store at –20°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection