Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Pre-formulated TEAB and 50% hydroxylamine designed for use in TMT labeling and quenching of peptides.
Description | 1M Triethylammonium bicarbonate (TEAB) for TMT experiments |
Product Type | Buffer |
Product Line | TMT™ |
Starting Material | Cell Lysate |
Workflow Step | Protein Labeling |
Quantity | 50 mL |
Unit Size | 50 mL |
Description | 50% Hydroxylamine for TMT experiments |
Product Type | Buffer |
Product Line | TMT™ |
Starting Material | Cell Lysate |
Workflow Step | Protein Labeling |
Quantity | 5 mL |
Unit Size | 5 mL |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
90114 | 50 mL | 1M Triethylammonium bicarbonate (TEAB) for TMT experiments | 141.65 Online price 154.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
90115 | 5 mL | 50% Hydroxylamine for TMT experiments | 96.65 Online price 103.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Triethylamonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.