Applied Biosystems™

PrimeView Human Genome U219 Array Plate

Catalog number: 901605
Have Questions?
Applied Biosystems™

PrimeView Human Genome U219 Array Plate

Catalog number: 901605
Have Questions?
Catalog Number
901605
Unit Size
1 x 16 array plate
Price (USD)
Catalog Number
Unit Size
1 x 24 array plate
Price (USD)
Catalog Number
Unit Size
1 x 96 array plate
Price (USD)
Catalog NumberUnit SizePrice (USD)
9016051 x 16 array plate
9016041 x 24 array plate
9015951 x 96 array plate
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Proven performance from the industry standard
The PrimeView Human Genome U219 96-array Plate and Trays enables high-throughput expression profiling of 96 samples at a time using probe sets with an emphasis on established, well-annotated content. Sequences used in the design of the array were selected from the UniGene database 219, RefSeq version 36, and full-length human mRNAs from GenBank™.

The Human Genome U219 Array enables you to:
• Measure gene expression of more than 36,000 transcripts and variants per sample
• Get accurate and reproducible gene expression data by using multiple independent measurements for each transcript

Key benefits
• Enables increased productivity and efficiency through parallel processing
• Process 16, 24, or 96 samples on a single array plate
• Offers excellent gene expression accuracy and reproducibility
• Multiple independent measurements per transcript for increased confidence in your results
   - 11 probes per set for well-annotated sequences
   - 9 probes per set for the remainder
• Provides complete coverage of the annotated genome

Content profile
• The Human Genome U219 Array is comprised of more than 530,000 probes covering more than 36,000 transcripts and variants, which represent more than 20,000 genes mapped through UniGene or via RefSeq annotation.
• The EST and mRNA sequences used in the design were clustered and assembled to create consensus sequences that represent alternative splice forms. Each assembly was then analyzed for orientation and alternative 3' end evidence.
• Content was chosen to cover all well-annotated genes and transcripts from RefSeq v36 and by leveraging all available EST and mRNA evidence that fall into the same clusters, to rigorously detect alternate 3' ends of those
well-annotated genes.
• More than 1,000 probe sets represent transcripts that have no official gene symbol in UniGene, but are based on predicted RefSeq sequences and UniGene clusters with good evidence of actual transcription.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Human Genome U219 Array Plate
Array
Transcriptome Profiling
Number of Arrays
16 arrays
Format
Array Plate and Trays
Species
Human
No. of Samples
96

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text