Applied Biosystems™
HT WT Expression Assay Consumables for Biomek FXp Target Prep Express
Catalog number: 901646
Have Questions?
Applied Biosystems™
HT WT Expression Assay Consumables for Biomek FXp Target Prep Express
Catalog number: 901646
Have Questions?
Catalog Number
901646
Unit Size
1 kit
Price (USD)
Catalog NumberUnit SizePrice (USD)
9016461 kit
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The HT WT Expression Assay Consumables is used with the WT Expression Kit for High-Throughput Robotics and the HT WT Terminal Labeling and Controls Kit for customers who use the Beckman Coulter Biomek™ FXp Target Prep Express Instrument (each sold separately).
The HT WT Terminal Labeling and Controls Kit contains the GeneChip™ HT WT Terminal Labeling Kit, GeneChip; Hybridization Control Kit, and GeneChipPoly-A Control Kit.
Key Features
• Low RNA input requirements — as little as 100 ng total RNA for a single round of amplification
• No separate rRNA depletion step
• Magnetic bead purification for enhanced recovery and ease of use
• StreamLined workflow for simplified protocol and reduced hands-on time
• Unbiased coverage of the transcriptome
The WT Expression Kit for High-Throughput Robotics includes:
• All reagents needed to generate sense cDNA ready for fragmentation and labeling
• Magnetic beads for enhanced purification efficiency
The HT WT Terminal Labeling and Controls Kit includes:
• WT Fragmentation and Labeling Kit required for target produced by the WT Expression Kit for High-Throughput Robotics
• Poly-A controls are added to input RNA to measure efficiency of target amplification by the WT Expression Kit for High-Throughput Robotics
• Hybridization controls that monitor the hybridization process for troubleshooting purposes
Related Links
HT WT Terminal Labeling and Controls Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Type
HT WT Expression Assay Consumable
For Use With (Equipment)
Biomek FXP Target Prep Express