Applied Biosystems™

HT WT Terminal Labeling and Controls Kit

Catalog number: 901648
Have Questions?
Applied Biosystems™

HT WT Terminal Labeling and Controls Kit

Catalog number: 901648
Have Questions?
Catalog Number
901648
Unit Size
1 x 96 reactions
Price (USD)
Catalog Number
Unit Size
1 x 24 reactions
Price (USD)
Catalog NumberUnit SizePrice (USD)
9016481 x 96 reactions
9016471 x 24 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The HT WT Terminal Labeling and Controls Kit is used with the WT Expression Kit for High-Throughput Robotics (sold separately). Both products can be combined with the HT WT Expression Assay Consumables for Biomek™ FXp Target Prep Express (sold separately) for customers who use the Beckman Coulter Biomek™ FXp Target Prep Express Instrument.

The HT WT Terminal Labeling and Controls Kit contains the GeneChip™ HT WT Terminal Labeling Kit, GeneChip™ Hybridization Control Kit, and GeneChip™ Poly-A Control Kit.

Key Features
• Low RNA input requirements — as little as 100 ng total RNA for a single round of amplification
• No separate rRNA depletion step
• Magnetic bead purification for enhanced recovery and ease of use
• StreamLined workflow for simplified protocol and reduced hands-on time
• Unbiased coverage of the transcriptome

The WT Expression Kit for High-Throughput Robotics includes:
• All reagents needed to generate sense cDNA ready for fragmentation and labeling
• Magnetic beads for enhanced purification efficiency

The HT WT Terminal Labeling and Controls Kit includes:
• WT Fragmentation and Labeling Kit required for target produced by the WT Expression Kit for High-Throughput Robotics
• Poly-A controls are added to input RNA to measure efficiency of target amplification by the WT Expression Kit for High-Throughput Robotics
• Hybridization controls that monitor the hybridization process for troubleshooting purposes

Related Links
HT WT Expression Assay Consumables for Biomek™ FXp Target Prep Express
HT WT Terminal Labeling and Controls Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
HT WT Terminal Labeling and Controls Kit
Detection Method
Biotin-based
Format
Kit
Final Product Type
DNA (Fragmented, Labeled)
Labeling Target
RNA (General)
Includes Label or Dye
Direct Labeling
Isolation Technology
Magnetic Bead
No. of Reactions
1 x 96
Product Type
WT Terminal Labeling Kit

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text