Applied Biosystems™

Axiom™ 2.0 Reagent Kit

Catalog number: 901758
Have Questions?
Applied Biosystems™

Axiom™ 2.0 Reagent Kit

Catalog number: 901758
Have Questions?
Catalog Number
901758
Unit Size
1 kit
Price (USD)
Catalog NumberUnit SizePrice (USD)
9017581 kitRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Axiom™ 2.0 Reagent Kits are designed for use with Axiom pre-designed and Axiom myDesign™ Arrays.

These kits are available in 96-, 384-, and 24- reaction formats. The kits include the reagents and control DNA required to produce hybridization-ready targets for Axiom pre-designed and Axiom myDesign Arrays. Axiom Reagent Kits can be used for genotyping of SNPs and indels on Axiom Genotyping Arrays and for the detection of microbial targets on Axiom Microbiome Arrays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
2.0 Reagent Kit
Format
96-, 384-, 24- Format
For Use With (Application)
Genotyping
Includes
Reagent, Control DNA
Reagent Type
Genotyping reagents

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text