See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Clinical Analyzers and Instruments
Microarray Kits
CytoScan™ Consumables Kit for NIMBUS™
Applied Biosystems™
CytoScan™ Consumables Kit for NIMBUS™
Catalog number:
902168
Related applications:
Microarray Analysis
Have Questions?
Contact Us
Applied Biosystems™
CytoScan™ Consumables Kit for NIMBUS™
Catalog number:
902168
Related applications:
Microarray Analysis
Have Questions?
Contact Us
Catalog Number
902168
Unit Size
1 kit
Price (USD)
Contact Us
Save to list
Catalog Number
Unit Size
Price (USD)
902168
1 kit
Contact Us
Save to list
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The CytoScan™ Consumables Kit for NIMBUS™ contains labware sufficient for 4x25 or 2x49 rxn runs, and is required for the NIMBUS Target Preparation Instrument (sold separately).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Type
Consumables Kit
For Use With (Equipment)
NIMBUS Target Preparation Instrument
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate