See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Molecular Biology Reagents and Kits
RNA Preparation and Purification
GeneTitan™ Hybridization Module for miRNA Plates
Applied Biosystems™
GeneTitan™ Hybridization Module for miRNA Plates
Catalog number:
902275
Related applications:
Microarray Analysis
Have Questions?
Contact Us
Applied Biosystems™
GeneTitan™ Hybridization Module for miRNA Plates
Catalog number:
902275
Related applications:
Microarray Analysis
Have Questions?
Contact Us
Catalog Number
902275
Unit Size
1 x 96 reactions
Price (USD)
1,086.00
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
902275
1 x 96 reactions
1,086.00
-
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The GeneTitan™ Hybridization Module for miRNA Plates is used with the miRNA 3.1 and 4.1 Array Plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)
miRNA Profiling
Product Type
Hybridization Module
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate