Applied Biosystems™

GeneChip™ Human Transcriptome Assay 2.0

Catalog number: 902310
Have Questions?
Applied Biosystems™

GeneChip™ Human Transcriptome Assay 2.0

Catalog number: 902310
Have Questions?
Catalog Number
902310
Unit Size
30 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
90231030 reactions
-
90230910 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Designed to empower next-generation expression profiling studies, the GeneChip™ Human Transcriptome Assay 2.0 provides the ability to go beyond gene-level expression profiling by providing the coverage and accuracy required to accurately detect all known transcript isoforms produced by a gene.

View the data sheet for details on HTA 2.0 content and coverage.

Comprehensive exploration of the transcriptome
Research has shown that the tens of thousands of human genes contain hundreds of thousands of exons, which produce hundreds of thousands of different transcript isoforms. These transcript isoforms are produced when the exons of a gene may be included within, or excluded from, the final, processed messenger RNA produced from that gene. Until now, measuring and analyzing these transcript isoforms has been nearly impossible due to technology limitations, sample input requirements, and lack of analysis capabilities/tools.

Comprehensive transcriptome analysis requires combining transcript diversity from multiple data sources
Most genes produce multiple transcript isoforms, and measuring changes in the relative abundance of each isoform provides new insights into disease and biology. HTA 2.0 has combined multiple data sources to ensure you are able to independently analyze the broadest collection of transcript isoforms available.

Data sources used to design and annotate the assay
RefSeq                                    Vertebrate Genome Annotation (Vega) database
Ensembl                                   MGC Mammalian Gene Collection (v10)
UCSC Known Genes               www.noncode.org
UCSC lincRNA transcripts        lncRNA db
Broad Institute, Human Body Map lincRNAs, and TUCP (transcripts of uncertain coding potential) catalog

Better data than 2 full lanes of sequencing
See the HTA 2.0 Flyer in the Documents Section below for additional information. HTA 2.0 provides superior accuracy and precision coupled with the most comprehensive view of the transcriptome.

Bioinformatics built into the array design; no assembly required
HTA 2.0 maximizes the amount of unique and valuable information possible by minimizing the conserved sequence synthesized on the array. This high-resolution array design contains an unprecedented >6.0 million probes covering coding transcripts and non-coding transcripts. 70% of the probes on this array cover exons for coding transcripts, and the remaining 30% of probes on the array cover exon-exon splice junctions and non-coding transcripts. The unparalleled coverage of this array provides the deepest insight into all coding and non-coding transcripts available.

Simple, fast, and free analysis solution
For the first time ever, HTA 2.0 coupled with Expression Console™ Software and TAC Software offers researchers a complete solution from data to decision making in minutes. This complete analysis solution is provided to all researchers using our expression arrays at no additional cost. In addition, HTA 2.0 data analysis is supported by the same analysis solutions and service providers being used for other expression array data.

Related Links
GeneChip™ Hybridization, Wash, and Stain Kit
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Human Transcriptome Assay 2.0
Array
Transcriptome Profiling
Number of Arrays
30 arrays
Format
Array Cartridge
Species
Human
Product Line
GeneChip™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text