GeneChip™ miRNA 4.1 Array Plate
GeneChip™ miRNA 4.1 Array Plate
Applied Biosystems™
GeneChip™ miRNA 4.1 Array Plate
Many diseases, including cancer, are frequently described as diseases of disordered gene expression. It is estimated that more than 30%Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90241096 arrays
902408TS16 arrays
90240924 arrays
Catalog number 902410
Price (USD)
-
Number of Arrays:
96 arrays
Many diseases, including cancer, are frequently described as diseases of disordered gene expression. It is estimated that more than 30% of protein translation of coding genes is regulated by miRNA. There is also a large amount of growing evidence suggesting miRNA interacts with long non-coding RNA in the signaling networks that regulate alternative splicing events, which impacts cellular processes such as apoptosis, proliferation, and differentiation. All of which have shown to be causative elements in diseases such as cancer.
Measuring the changes in these critical nodes of regulation is extremely important for deciphering the biological context of differentially expressed genes. This array design is a powerful tool for studying the role of small non-coding RNAs and their involvement in a broad spectrum of developmental and physiological mechanisms.
To keep pace with the discovery of new and novel miRNA, we are pleased to offer the miRNA 4.1 96-Array Plates and Trays in our catalog of miRNA arrays. This product offers updated content with the same high performance as the previous generation array.
miRNA 4.1 Array Plates and Trays help bring you closer to biology with:
Comprehensive coverage – Designed to interrogate all mature miRNA sequences in miRBase Release 20
Easily correlate miRNA results – Analysis files contain host gene ID, predicted and validated miRNA target genes, and clustered miRNA information
Easy analysis – Analyze human, mouse, rat, or every miRNA for all species using the same array
Low sample input – Requires as little as 130 ng total RNA
Simple, fast, and free analysis solution – Coupled with Expression Console™ Software and Transcriptome Analysis Console (TAC) Software, researchers have a complete solution from data to decision-making in minutes
Related Links
GeneTitan™ Wash Buffers A and B Module
B2 Control Oligo (3nM)
Genetitan™ miRNA Array Holding Buffer
FlashTag™ Biotin HSR RNA Labeling Kits
FlashTag™ Biotin HSR RNA Labeling Kits
GeneTitan™ Hybridization Module for miRNA Plates
GeneChip™ Hybridization Control Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
TypemiRNA 4.1 Array Plate
ArraymiRNA Profiling
Number of Arrays96 arrays
Format96-array Plate, Trays
SpeciesAll Organisms, Human, Mouse, Rat
Product LineGeneChip™
Quantity1 x 96-array plate
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link