Applied Biosystems™

GeneChip™ miRNA 4.0 Assay

Catalog number: 902446
Have Questions?
Applied Biosystems™

GeneChip™ miRNA 4.0 Assay

Catalog number: 902446
Have Questions?
Catalog Number
902446
Unit Size
30 samples
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 samples
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
90244630 samples
-
90244510 samples
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Many diseases, including cancer, are frequently described as diseases of disordered gene expression. It is estimated that more than 30% of protein translation of coding genes is regulated by miRNA. There is also a large amount of growing evidence suggesting miRNA interacts with long non-coding RNA in the signaling networks that regulate alternative splicing events, which impacts cellular processes such as apoptosis, proliferation, and differentiation– all of which have shown to be causative elements in diseases such as cancer.

Measuring the changes in these critical nodes of regulation is extremely important for deciphering the biological context of differentially expressed genes. This array design is a powerful tool for studying the role of small non-coding RNAs and their involvement in a broad spectrum of developmental and physiological mechanisms.

To keep pace with the discovery of new and novel miRNA, we are pleased to offer the GeneChip™ miRNA 4.0 Array and Flashtag™ Bundle to our growing catalog of miRNA arrays. This array offers updated content with the same high performance as the previous-generation array.

GeneChip miRNA 4.0 Arrays help bring you closer to biology with:
Comprehensive coverage – Designed to interrogate all mature miRNA sequences in miRBase Release 20
Easily correlate miRNA results – Analysis files contain host gene ID, predicted and validated miRNA target genes, and clustered miRNA information
Easy analysis – Analyze human, mouse, rat, or every miRNA for all species using the same array
Low sample input – Requires as little as 130 ng total RNA
Simple, fast, and free analysis solution – Coupled with Expression Console™ Software and Transcriptome Analysis Console (TAC) Software, researchers have a complete solution from data to decision-making in minutes

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Array Cartridge
Species
All
Number of Arrays
30
Product Line
GeneChip™
Product Type
miRNA 4.0 Assay

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text