Applied Biosystems™

UK Biobank Axiom™ Array

Catalog number: 902502
Have Questions?
Applied Biosystems™

UK Biobank Axiom™ Array

Catalog number: 902502
Have Questions?
Catalog Number
902502
Unit Size
1 plate
Price (USD)
Catalog NumberUnit SizePrice (USD)
9025021 plate
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
UK Biobank Axiom™ Array was designed by and for thought leaders in the human genetics community for high-throughput, high-value genotyping of large sample cohorts to explore the genetics of complex diseases and translational research with a single comprehensive low-cost solution. This array contains one 96-array plate that allows for genome-wide genotyping of large sample collections such as those screened at biobanks, genome centers, and core labs.

GWAS
• Intelligent marker selection enables imputation of millions of additional SNPs
• The genomic content module of each array is optimized for specific populations and is customizable to any 1000 Genomes population study by adding additional markers of your choosing

Pharmacogenomic markers
• Markers were selected to represent phases of absorption, distribution, metabolism, and excretion (ADME)

Inflammation and HLA
• Contains markers with evidence for association with autoimmune and inflammation
• Covers variants in genes in the HLA and KIR regions known to be important in immune response

Exome
• Rare, non-synonymous coding SNPs and indels in protein coding regions of the genome
• A majority of variants are rare with minor allele frequency (MAF) <1%
• Newly discovered loss-of-function content
• SNPs and indels identified from a sequencing initiative of 26,000 individuals
• Known disease-causing mutations

eQTLs
• Markers that have known associations to RNA expression traits
• Captures unique eQTLs contained in the NCBI Genotype-Tissue Expression (GTEx) eQTL database, GEUVIDAS project, and several other discovery projects

Human disease
• Alzheimer-s disease, including coverage of ApoE
• Blood phenotypes
• Cancer common variants
• Cardiometabolic

Choose your content
• Order the pre-designed array and customize it with markers of your choice
• Custom markers may be de novo or selected from the Axiom Genomic Database of genotype-tested markers

Please note: Reagent kits do not include plastic consumables required to run the assay on the Beckman Biomek™ FXP Target Prep Express System. Please contact us for a list of required plastic consumables.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Array
Genotyping
Number of Arrays
96 arrays
Format
96-array Plate
Species
Human
For Use With (Application)
Microarray Analysis

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text