Applied Biosystems™

Axiom™ Microbiome, 24 array plate

Catalog number: 902903
Have Questions?
Applied Biosystems™

Axiom™ Microbiome, 24 array plate

Catalog number: 902903
Have Questions?
Catalog Number
902903
Unit Size
1 plate
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
9029031 plate
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Easy, streamlined solution for microbial profiling
Axiom Microbiome Array enables the detection of all known microorganisms in a sample with species- and strain-level identification in a single, scalable assay. Utilizing the robust biochemistry of Axiom ™ 2.0 Assay coupled with manual and automated target preparation methods, this streamlined protocol provides consistent and high-quality results. A simplified, upstream reverse transcriptase reaction enables the detection of RNA virus genomes. Hands-free array processing is performed on GeneTitan Multi-Channel Instrument. The complete solution includes arrays, reagent kits, and free, easy-to-use microbiome analysis software (see Figures 1 and 2).

Highlights:
• Expert design for profiling human and animal microbiomes with a single assay
•Detects greater diversity and higher resolution than 16s rRNA gene sequencing
•Allows genetic profiling of all organisms present in a sample
•Comprehensive coverage of over 11,000 organisms and five microbial domains: archaea, bacteria, fungi, protozoa, and viruses
• Allows identification to the species, strain and sequence level.
•Flexible 24- and 96-array plate formats
•Sample type agnostic: applications in nutrigenomics, agrigenomics, and animal research
•Simplified microbial detection analysis

See also Axiom Microbiome, 96 array plate and Axiom Microbial Detection Analysis Software (MiDAS).

See Figure 2 for target categories represented on Axiom Microbiome Arrays.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Array
Genotyping
Number of Arrays
24 arrays
Format
24-array Plate
Species
Protozoa, Archaea, Bacteria, Fungi, Virus
For Use With (Application)
Microarray Analysis

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text