Applied Biosystems™

Clariom™ D Pico Assay, human

Catalog number: 902925
Have Questions?
Applied Biosystems™

Clariom™ D Pico Assay, human

Catalog number: 902925
Have Questions?
Catalog Number
902925
Unit Size
30 reactions
Price (USD)
Catalog Number
Unit Size
12 reactions
Price (USD)
Catalog NumberUnit SizePrice (USD)
90292530 reactions
90292412 reactions
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Accelerate your biomarker discovery from deep within the transcriptome with Clariom D Pico Assays for human samples, the next generation of transcriptome-level expression profiling tools. Human Clariom D Pico Assays provide a highly detailed view of the transcriptome and offer the fastest path to the results you need for your research. Clariom D Pico Assays allow translational research scientists to generate high-fidelity biomarker signatures quickly and easily. Based on industry-leading microarray technology, the novel Clariom D Pico Assay design provides the most intricate transcriptome-wide gene- and exon-level expression profiles, including the ability to detect alternative splicing events of coding and long non-coding (lnc)RNA, in a single three-day experiment.

Expand your potential to discover novel, informative biomarkers
The number of known transcribed genes has expanded rapidly in recent years, providing more sources for actionable biomarkers, such as transcript variants and lncRNA, that can be used for clinical utility and advancing our understanding of disease mechanisms. Such biomarkers can be missed by lengthy, complex, and costly sequencing and targeted expression approaches, leading to irreproducible signatures and wasted time and money.

With full coverage of the transcribed genome including all known coding and non-coding splice variants, compatibility with clinical sample types, and flexible data analysis software, Clariom D Pico assays are the premier tools for translational researchers performing complex expression biomarker discovery studies and wanting the fastest path to robust, clinically relevant, and actionable results.

Get all the data you need
• Rapidly identify complex disease signatures from >540,000 transcripts sourced from the largest number of public databases, the most comprehensive coverage of the human transcriptome available, to ensure biomarkers are not missed.
• Confidently detect genes, exons, and alternative splicing events that give rise to coding RNA and lncRNA isoforms.
• Detect rare and low-expressing transcripts otherwise not detected by common sequencing approaches.
• Go from data to insight in minutes with intuitive, highly visual, free analysis software.

When you have precious samples, get it right the first time.
• Generate robust expression profiles from as little as 100 pg of total RNA—as few as 10 cells.
• Utilize RNA from various sample types including blood, cells, and fresh/fresh-frozen or FFPE tissues.
• Preserve sample integrity and reduce data variability with an assay that does not require a globin or rRNA removal step.

Clariom D solutions are available in a single sample (cartridge array) format for use on the GeneChip™ 3000 instrument system and include reagents and fast, simple Transcriptome Analysis Console (TAC) software to analyze and visualize global expression patterns of genes, exons, pathways, and alternative splicing events.

Get the coverage you require, the reproducibility you need, and the insights you want to act on your discoveries.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
D Pico Assay
Array
Transcriptome Profiling
Number of Arrays
30 arrays
Format
Array Cartridge
Species
Human
Product Line
Applied Biosystems™
Shipping Condition
Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & Storage

• Control HeLA RNA, store at -20°C
• Hybridization Control Kit, store at -20°C
• WT Pico Amplification Kit, Module 1, store at -20°C
• WT Pico Amplification Kit, Module 2, store at -20°C
• WT Pico Amplification Kit, Module 3, store at 4°C
• Clariom D 30 Arrays, human, store at 4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text